Assay Details
| Gene Symbol: CYR61 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK096420.1 | - | 1 | 519 | 159 | |
| AL832891.1 | - | 1 | 337 | 159 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

CYR61| Gene Symbol: CYR61 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK096420.1 | - | 1 | 519 | 159 | |
| AL832891.1 | - | 1 | 337 | 159 | ||
Set Membership: |
|