| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100015599.hg.1 |
ENST00000336751.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100015599.hg.1 |
ENST00000336751.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100015599.hg.1 |
ENST00000336751.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100015599.hg.1 |
ENST00000336751.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100015599.hg.1 |
ENST00000336751.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100015599.hg.1 |
ENST00000336751.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100015599.hg.1 |
ENST00000336751.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100015599.hg.1 |
ENST00000336751.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100015599.hg.1 |
ENST00000336751.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100015599.hg.1 |
ENST00000336751.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100015599.hg.1 |
ENST00000357836.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100015599.hg.1 |
ENST00000357836.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100015599.hg.1 |
ENST00000357836.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100015599.hg.1 |
ENST00000357836.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100015599.hg.1 |
ENST00000357836.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100015599.hg.1 |
ENST00000357836.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100015599.hg.1 |
ENST00000357836.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100015599.hg.1 |
ENST00000357836.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100015599.hg.1 |
ENST00000357836.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100015599.hg.1 |
ENST00000357836.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100015599.hg.1 |
ENST00000421822.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100015599.hg.1 |
ENST00000421822.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100015599.hg.1 |
ENST00000421822.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100015599.hg.1 |
ENST00000421822.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100015599.hg.1 |
ENST00000421822.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100015599.hg.1 |
ENST00000421822.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100015599.hg.1 |
ENST00000475797.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100015599.hg.1 |
ENST00000475797.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100015599.hg.1 |
ENST00000475797.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100015599.hg.1 |
ENST00000475797.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100015599.hg.1 |
ENST00000497365.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100015599.hg.1 |
ENST00000497365.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100015599.hg.1 |
ENST00000497365.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100015599.hg.1 |
ENST00000497365.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100015599.hg.1 |
ENST00000497365.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100015599.hg.1 |
NM_145277.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100015599.hg.1 |
NM_145277.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100015599.hg.1 |
NM_145277.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100015599.hg.1 |
NM_145277.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100015599.hg.1 |
NM_145277.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100015599.hg.1 |
NM_145277.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100015599.hg.1 |
NM_145277.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100015599.hg.1 |
NM_145277.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100015599.hg.1 |
NM_145277.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100015599.hg.1 |
NM_202004.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100015599.hg.1 |
NM_202004.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100015599.hg.1 |
NM_202004.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100015599.hg.1 |
NM_202004.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100015599.hg.1 |
NM_202004.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100015599.hg.1 |
NM_202004.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100015599.hg.1 |
NM_213652.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100015599.hg.1 |
NM_213652.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100015599.hg.1 |
NM_213652.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100015599.hg.1 |
NM_213652.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100015599.hg.1 |
NM_213652.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100015599.hg.1 |
NM_213653.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100015599.hg.1 |
NM_213653.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100015599.hg.1 |
NM_213653.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100015599.hg.1 |
NM_213653.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100015599.hg.1 |
NM_213653.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100015599.hg.1 |
NM_213653.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100015599.hg.1 |
NM_213653.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100015599.hg.1 |
NM_213653.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100015599.hg.1 |
NM_213653.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100015599.hg.1 |
NM_213653.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100015599.hg.1 |
NM_213653.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100015599.hg.1 |
ENST00000336751.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100015599.hg.1 |
ENST00000336751.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100015599.hg.1 |
ENST00000336751.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100015599.hg.1 |
ENST00000336751.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100015599.hg.1 |
ENST00000336751.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100015599.hg.1 |
ENST00000336751.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100015599.hg.1 |
ENST00000336751.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100015599.hg.1 |
ENST00000336751.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100015599.hg.1 |
ENST00000336751.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100015599.hg.1 |
ENST00000336751.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100015599.hg.1 |
ENST00000357836.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100015599.hg.1 |
ENST00000357836.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100015599.hg.1 |
ENST00000357836.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100015599.hg.1 |
ENST00000357836.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100015599.hg.1 |
ENST00000357836.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100015599.hg.1 |
ENST00000357836.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100015599.hg.1 |
ENST00000357836.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100015599.hg.1 |
ENST00000357836.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100015599.hg.1 |
ENST00000357836.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100015599.hg.1 |
ENST00000357836.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100015599.hg.1 |
ENST00000421822.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100015599.hg.1 |
ENST00000421822.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100015599.hg.1 |
ENST00000421822.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100015599.hg.1 |
ENST00000421822.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100015599.hg.1 |
ENST00000421822.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100015599.hg.1 |
ENST00000421822.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100015599.hg.1 |
ENST00000475797.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100015599.hg.1 |
ENST00000475797.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100015599.hg.1 |
ENST00000475797.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100015599.hg.1 |
ENST00000475797.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100015599.hg.1 |
ENST00000497365.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100015599.hg.1 |
ENST00000497365.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100015599.hg.1 |
ENST00000497365.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100015599.hg.1 |
ENST00000497365.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100015599.hg.1 |
ENST00000497365.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100015599.hg.1 |
NM_145277.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100015599.hg.1 |
NM_145277.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100015599.hg.1 |
NM_145277.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100015599.hg.1 |
NM_145277.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100015599.hg.1 |
NM_145277.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100015599.hg.1 |
NM_145277.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100015599.hg.1 |
NM_145277.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100015599.hg.1 |
NM_145277.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100015599.hg.1 |
NM_145277.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100015599.hg.1 |
NM_202004.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100015599.hg.1 |
NM_202004.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100015599.hg.1 |
NM_202004.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100015599.hg.1 |
NM_202004.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100015599.hg.1 |
NM_202004.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100015599.hg.1 |
NM_202004.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100015599.hg.1 |
NM_213652.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100015599.hg.1 |
NM_213652.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100015599.hg.1 |
NM_213652.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100015599.hg.1 |
NM_213652.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100015599.hg.1 |
NM_213652.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100015599.hg.1 |
NM_213653.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100015599.hg.1 |
NM_213653.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100015599.hg.1 |
NM_213653.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100015599.hg.1 |
NM_213653.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100015599.hg.1 |
NM_213653.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100015599.hg.1 |
NM_213653.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100015599.hg.1 |
NM_213653.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100015599.hg.1 |
NM_213653.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100015599.hg.1 |
NM_213653.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100015599.hg.1 |
NM_213653.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100015599.hg.1 |
NM_213653.3 |
Refseq |