Assay Details
| Gene Symbol: RASSF5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_182663.3 | NP_872604.1 | 6 | 2559 | 97 | |
| NM_182664.3 | NP_872605.1 | 5 | 2443 | 97 | ||
| NM_182665.3 | NP_872606.1 | 5 | 2437 | 97 | ||
| GenBank mRNA | AB209145.1 | - | 4 | 4519 | 97 | |
| AF445801.1 | - | 5 | 2437 | 97 | ||
| AK056568.1 | - | 2 | 2203 | 97 | ||
| AK074115.1 | - | 3 | 3434 | 97 | ||
| AL832784.1 | - | 5 | 2337 | 97 | ||
| BC004270.1 | - | 7 | 2413 | 3613993 | 97 | |
| BC007203.1 | - | 7 | 2413 | 2967057 | 97 | |
| BC042651.1 | - | 5 | 2417 | 4342962 | 97 | |
More Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0100011378.hg.1 | ENST00000577571.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011378.hg.1 | ENST00000577571.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011378.hg.1 | ENST00000577571.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011378.hg.1 | ENST00000577571.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011378.hg.1 | ENST00000577571.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011378.hg.1 | ENST00000577571.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011378.hg.1 | ENST00000577571.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011378.hg.1 | ENST00000577571.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011378.hg.1 | ENST00000577571.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011378.hg.1 | ENST00000579436.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011378.hg.1 | ENST00000579436.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011378.hg.1 | ENST00000579436.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011378.hg.1 | ENST00000579436.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011378.hg.1 | ENST00000579436.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011378.hg.1 | ENST00000579436.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011378.hg.1 | ENST00000579436.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011378.hg.1 | ENST00000579436.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011378.hg.1 | ENST00000579436.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011378.hg.1 | ENST00000579942.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011378.hg.1 | ENST00000579942.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011378.hg.1 | ENST00000579942.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011378.hg.1 | ENST00000579942.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011378.hg.1 | ENST00000579942.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011378.hg.1 | ENST00000579942.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011378.hg.1 | ENST00000580449.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011378.hg.1 | ENST00000580449.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011378.hg.1 | ENST00000580449.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011378.hg.1 | ENST00000580449.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011378.hg.1 | ENST00000580449.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011378.hg.1 | ENST00000580449.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011378.hg.1 | ENST00000580449.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011378.hg.1 | ENST00000580449.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011378.hg.1 | ENST00000581503.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011378.hg.1 | ENST00000581503.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011378.hg.1 | ENST00000581503.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011378.hg.1 | ENST00000581503.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011378.hg.1 | ENST00000581503.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011378.hg.1 | ENST00000581503.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011378.hg.1 | ENST00000581503.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011378.hg.1 | ENST00000581503.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011378.hg.1 | ENST00000581503.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011378.hg.1 | ENST00000581503.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011378.hg.1 | ENST00000581503.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011378.hg.1 | ENST00000581888.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011378.hg.1 | ENST00000581888.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011378.hg.1 | ENST00000581888.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011378.hg.1 | ENST00000581888.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011378.hg.1 | ENST00000581888.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011378.hg.1 | ENST00000581888.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011378.hg.1 | ENST00000581888.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011378.hg.1 | ENST00000581888.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011378.hg.1 | ENST00000605120.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011378.hg.1 | ENST00000605120.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011378.hg.1 | ENST00000605120.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011378.hg.1 | ENST00000605653.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011378.hg.1 | ENST00000605653.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011378.hg.1 | ENST00000605653.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011378.hg.1 | ENST00000605653.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011378.hg.1 | NM_182663.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011378.hg.1 | NM_182663.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011378.hg.1 | NM_182663.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011378.hg.1 | NM_182663.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011378.hg.1 | NM_182663.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011378.hg.1 | NM_182663.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011378.hg.1 | NM_182663.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011378.hg.1 | NM_182663.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011378.hg.1 | NM_182663.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011378.hg.1 | NM_182663.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011378.hg.1 | NM_182664.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011378.hg.1 | NM_182664.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011378.hg.1 | NM_182664.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011378.hg.1 | NM_182664.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011378.hg.1 | NM_182664.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011378.hg.1 | NM_182664.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011378.hg.1 | NM_182664.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011378.hg.1 | NM_182664.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011378.hg.1 | NM_182664.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011378.hg.1 | NM_182665.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011378.hg.1 | NM_182665.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011378.hg.1 | NM_182665.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011378.hg.1 | NM_182665.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011378.hg.1 | NM_182665.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011378.hg.1 | NM_182665.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011378.hg.1 | NM_182665.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011378.hg.1 | NM_182665.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011378.hg.1 | NM_182665.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011378.hg.1 | ENST00000577571.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011378.hg.1 | ENST00000577571.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011378.hg.1 | ENST00000577571.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011378.hg.1 | ENST00000577571.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011378.hg.1 | ENST00000577571.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011378.hg.1 | ENST00000577571.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011378.hg.1 | ENST00000577571.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011378.hg.1 | ENST00000577571.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011378.hg.1 | ENST00000577571.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011378.hg.1 | ENST00000579436.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011378.hg.1 | ENST00000579436.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011378.hg.1 | ENST00000579436.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011378.hg.1 | ENST00000579436.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011378.hg.1 | ENST00000579436.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011378.hg.1 | ENST00000579436.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011378.hg.1 | ENST00000579436.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011378.hg.1 | ENST00000579436.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011378.hg.1 | ENST00000579436.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011378.hg.1 | ENST00000579942.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011378.hg.1 | ENST00000579942.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011378.hg.1 | ENST00000579942.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011378.hg.1 | ENST00000579942.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011378.hg.1 | ENST00000579942.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011378.hg.1 | ENST00000579942.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011378.hg.1 | ENST00000580449.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011378.hg.1 | ENST00000580449.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011378.hg.1 | ENST00000580449.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011378.hg.1 | ENST00000580449.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011378.hg.1 | ENST00000580449.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011378.hg.1 | ENST00000580449.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011378.hg.1 | ENST00000580449.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011378.hg.1 | ENST00000580449.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011378.hg.1 | ENST00000581503.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011378.hg.1 | ENST00000581503.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011378.hg.1 | ENST00000581503.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011378.hg.1 | ENST00000581503.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011378.hg.1 | ENST00000581503.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011378.hg.1 | ENST00000581503.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011378.hg.1 | ENST00000581503.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011378.hg.1 | ENST00000581503.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011378.hg.1 | ENST00000581503.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011378.hg.1 | ENST00000581503.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011378.hg.1 | ENST00000581503.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011378.hg.1 | ENST00000581888.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011378.hg.1 | ENST00000581888.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011378.hg.1 | ENST00000581888.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011378.hg.1 | ENST00000581888.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011378.hg.1 | ENST00000581888.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011378.hg.1 | ENST00000581888.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011378.hg.1 | ENST00000581888.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011378.hg.1 | ENST00000581888.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011378.hg.1 | ENST00000605120.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011378.hg.1 | ENST00000605120.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011378.hg.1 | ENST00000605120.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011378.hg.1 | ENST00000605653.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011378.hg.1 | ENST00000605653.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011378.hg.1 | ENST00000605653.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011378.hg.1 | ENST00000605653.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011378.hg.1 | NM_182663.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011378.hg.1 | NM_182663.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011378.hg.1 | NM_182663.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011378.hg.1 | NM_182663.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011378.hg.1 | NM_182663.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011378.hg.1 | NM_182663.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011378.hg.1 | NM_182663.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011378.hg.1 | NM_182663.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011378.hg.1 | NM_182663.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011378.hg.1 | NM_182663.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011378.hg.1 | NM_182664.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011378.hg.1 | NM_182664.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011378.hg.1 | NM_182664.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011378.hg.1 | NM_182664.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011378.hg.1 | NM_182664.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011378.hg.1 | NM_182664.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011378.hg.1 | NM_182664.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011378.hg.1 | NM_182664.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011378.hg.1 | NM_182664.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011378.hg.1 | NM_182665.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011378.hg.1 | NM_182665.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011378.hg.1 | NM_182665.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011378.hg.1 | NM_182665.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011378.hg.1 | NM_182665.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011378.hg.1 | NM_182665.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011378.hg.1 | NM_182665.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011378.hg.1 | NM_182665.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011378.hg.1 | NM_182665.3 | Refseq |

RASSF5
Genomic Map