Assay Details
| Gene Symbol: BMS1P6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | BC008503.1 | - | 1 | 717 | 123 | |
More Information
Set Membership: |
|
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1000010555.hg.1 | ENST00000413193.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010555.hg.1 | ENST00000413193.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010555.hg.1 | ENST00000413193.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010555.hg.1 | ENST00000413193.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010555.hg.1 | ENST00000413193.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010555.hg.1 | ENST00000413193.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010555.hg.1 | ENST00000413193.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010555.hg.1 | ENST00000413193.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010555.hg.1 | ENST00000413193.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010555.hg.1 | ENST00000413193.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010555.hg.1 | ENST00000431840.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010555.hg.1 | ENST00000431840.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010555.hg.1 | ENST00000431840.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010555.hg.1 | ENST00000431840.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010555.hg.1 | ENST00000431840.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010555.hg.1 | ENST00000431840.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010555.hg.1 | ENST00000431840.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010555.hg.1 | ENST00000431840.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010555.hg.1 | ENST00000431840.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010555.hg.1 | ENST00000431840.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010555.hg.1 | ENST00000450360.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010555.hg.1 | ENST00000450360.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010555.hg.1 | ENST00000450360.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010555.hg.1 | ENST00000450360.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010555.hg.1 | ENST00000450360.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010555.hg.1 | ENST00000450360.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010555.hg.1 | ENST00000450360.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010555.hg.1 | ENST00000450360.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010555.hg.1 | ENST00000450360.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010555.hg.1 | ENST00000450360.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010555.hg.1 | ENST00000452145.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010555.hg.1 | ENST00000452145.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010555.hg.1 | ENST00000452145.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010555.hg.1 | ENST00000452145.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010555.hg.1 | ENST00000452145.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010555.hg.1 | ENST00000452145.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010555.hg.1 | ENST00000452145.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010555.hg.1 | ENST00000452145.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010555.hg.1 | ENST00000452145.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010555.hg.1 | ENST00000452145.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010555.hg.1 | ENST00000452145.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010555.hg.1 | ENST00000452145.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010555.hg.1 | ENST00000532854.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010555.hg.1 | ENST00000532854.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010555.hg.1 | ENST00000532854.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010555.hg.1 | ENST00000532854.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010555.hg.1 | ENST00000532854.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010555.hg.1 | ENST00000532854.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010555.hg.1 | ENST00000532854.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010555.hg.1 | ENST00000532854.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010555.hg.1 | ENST00000532854.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010555.hg.1 | ENST00000532854.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010555.hg.1 | ENST00000605970.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010555.hg.1 | ENST00000605970.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010555.hg.1 | ENST00000605970.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010555.hg.1 | ENST00000605970.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010555.hg.1 | ENST00000605970.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010555.hg.1 | ENST00000605970.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010555.hg.1 | ENST00000605970.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010555.hg.1 | ENST00000605970.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010555.hg.1 | ENST00000605970.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010555.hg.1 | ENST00000605970.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010555.hg.1 | ENST00000605970.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010555.hg.1 | ENST00000605970.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010555.hg.1 | ENST00000605970.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010555.hg.1 | ENST00000605970.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010555.hg.1 | ENST00000605970.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010555.hg.1 | NM_001190810.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010555.hg.1 | NM_001190810.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010555.hg.1 | NM_001190810.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010555.hg.1 | NM_001190810.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010555.hg.1 | NM_001190810.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010555.hg.1 | NM_001190810.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010555.hg.1 | NM_001190810.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010555.hg.1 | NM_001190810.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010555.hg.1 | NM_001190810.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010555.hg.1 | NM_001190810.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010555.hg.1 | NM_001190810.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010555.hg.1 | NM_001190810.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010555.hg.1 | NR_024495.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010555.hg.1 | NR_024495.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010555.hg.1 | NR_024495.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010555.hg.1 | NR_024495.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010555.hg.1 | NR_024495.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010555.hg.1 | NR_024495.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010555.hg.1 | NR_024495.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010555.hg.1 | NR_024495.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010555.hg.1 | NR_024495.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010555.hg.1 | NR_024495.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010555.hg.1 | NR_024495.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000010555.hg.1 | ENST00000413193.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000010555.hg.1 | ENST00000413193.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000010555.hg.1 | ENST00000413193.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000010555.hg.1 | ENST00000413193.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000010555.hg.1 | ENST00000413193.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000010555.hg.1 | ENST00000413193.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000010555.hg.1 | ENST00000413193.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000010555.hg.1 | ENST00000413193.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000010555.hg.1 | ENST00000413193.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000010555.hg.1 | ENST00000413193.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000010555.hg.1 | ENST00000431840.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000010555.hg.1 | ENST00000431840.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000010555.hg.1 | ENST00000431840.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000010555.hg.1 | ENST00000431840.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000010555.hg.1 | ENST00000431840.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000010555.hg.1 | ENST00000431840.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000010555.hg.1 | ENST00000431840.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000010555.hg.1 | ENST00000431840.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000010555.hg.1 | ENST00000431840.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000010555.hg.1 | ENST00000431840.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000010555.hg.1 | ENST00000450360.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000010555.hg.1 | ENST00000450360.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000010555.hg.1 | ENST00000450360.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000010555.hg.1 | ENST00000450360.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000010555.hg.1 | ENST00000450360.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000010555.hg.1 | ENST00000450360.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000010555.hg.1 | ENST00000450360.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000010555.hg.1 | ENST00000450360.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000010555.hg.1 | ENST00000450360.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000010555.hg.1 | ENST00000450360.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000010555.hg.1 | ENST00000452145.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000010555.hg.1 | ENST00000452145.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000010555.hg.1 | ENST00000452145.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000010555.hg.1 | ENST00000452145.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000010555.hg.1 | ENST00000452145.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000010555.hg.1 | ENST00000452145.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000010555.hg.1 | ENST00000452145.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000010555.hg.1 | ENST00000452145.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000010555.hg.1 | ENST00000452145.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000010555.hg.1 | ENST00000452145.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000010555.hg.1 | ENST00000452145.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000010555.hg.1 | ENST00000452145.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000010555.hg.1 | ENST00000532854.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000010555.hg.1 | ENST00000532854.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000010555.hg.1 | ENST00000532854.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000010555.hg.1 | ENST00000532854.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000010555.hg.1 | ENST00000532854.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000010555.hg.1 | ENST00000532854.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000010555.hg.1 | ENST00000532854.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000010555.hg.1 | ENST00000532854.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000010555.hg.1 | ENST00000532854.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000010555.hg.1 | ENST00000532854.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000010555.hg.1 | ENST00000605970.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000010555.hg.1 | ENST00000605970.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000010555.hg.1 | ENST00000605970.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000010555.hg.1 | ENST00000605970.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000010555.hg.1 | ENST00000605970.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000010555.hg.1 | ENST00000605970.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000010555.hg.1 | ENST00000605970.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000010555.hg.1 | ENST00000605970.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000010555.hg.1 | ENST00000605970.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000010555.hg.1 | ENST00000605970.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000010555.hg.1 | ENST00000605970.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000010555.hg.1 | ENST00000605970.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000010555.hg.1 | ENST00000605970.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000010555.hg.1 | ENST00000605970.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000010555.hg.1 | ENST00000605970.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000010555.hg.1 | NM_001190810.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000010555.hg.1 | NM_001190810.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000010555.hg.1 | NM_001190810.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000010555.hg.1 | NM_001190810.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000010555.hg.1 | NM_001190810.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000010555.hg.1 | NM_001190810.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000010555.hg.1 | NM_001190810.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000010555.hg.1 | NM_001190810.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000010555.hg.1 | NM_001190810.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000010555.hg.1 | NM_001190810.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000010555.hg.1 | NM_001190810.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000010555.hg.1 | NM_001190810.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000010555.hg.1 | NR_024495.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000010555.hg.1 | NR_024495.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000010555.hg.1 | NR_024495.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000010555.hg.1 | NR_024495.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000010555.hg.1 | NR_024495.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000010555.hg.1 | NR_024495.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000010555.hg.1 | NR_024495.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000010555.hg.1 | NR_024495.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000010555.hg.1 | NR_024495.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000010555.hg.1 | NR_024495.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000010555.hg.1 | NR_024495.2 | Refseq |

BMS1P6
Genomic Map