Assay Details
| Gene Symbol: SKINT1L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_026749.2 | 3 - 4 | 193 | 132 | ||
| GenBank mRNA | DN994130.1 | - | 3 - 4 | 201 | 132 | |
More Information
Set Membership: |
|
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0100014117.hg.1 | ENST00000401057.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014117.hg.1 | ENST00000401057.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014117.hg.1 | ENST00000401057.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014117.hg.1 | ENST00000401057.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014117.hg.1 | ENST00000401057.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014117.hg.1 | ENST00000401057.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014117.hg.1 | ENST00000401057.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014117.hg.1 | ENST00000452503.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014117.hg.1 | ENST00000452503.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014117.hg.1 | ENST00000452503.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014117.hg.1 | ENST00000452503.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014117.hg.1 | ENST00000452503.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014117.hg.1 | ENST00000452503.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014117.hg.1 | ENST00000453388.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014117.hg.1 | ENST00000453388.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014117.hg.1 | ENST00000453388.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014117.hg.1 | ENST00000453388.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014117.hg.1 | ENST00000453388.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014117.hg.1 | ENST00000604015.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014117.hg.1 | ENST00000604015.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014117.hg.1 | NR_026749.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014117.hg.1 | NR_026749.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014117.hg.1 | NR_026749.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014117.hg.1 | NR_026749.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014117.hg.1 | NR_026749.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014117.hg.1 | NR_026749.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014117.hg.1 | NR_026749.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014117.hg.1 | NR_026749.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014117.hg.1 | NR_026749.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014117.hg.1 | NR_026749.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014117.hg.1 | NR_026749.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014117.hg.1 | NR_026749.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014117.hg.1 | NR_026749.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014117.hg.1 | NR_026749.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100014117.hg.1 | ENST00000401057.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014117.hg.1 | ENST00000401057.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014117.hg.1 | ENST00000401057.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014117.hg.1 | ENST00000401057.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014117.hg.1 | ENST00000401057.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014117.hg.1 | ENST00000401057.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014117.hg.1 | ENST00000401057.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014117.hg.1 | ENST00000452503.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014117.hg.1 | ENST00000452503.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014117.hg.1 | ENST00000452503.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014117.hg.1 | ENST00000452503.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014117.hg.1 | ENST00000452503.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014117.hg.1 | ENST00000452503.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014117.hg.1 | ENST00000453388.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014117.hg.1 | ENST00000453388.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014117.hg.1 | ENST00000453388.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014117.hg.1 | ENST00000453388.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014117.hg.1 | ENST00000453388.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014117.hg.1 | ENST00000604015.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014117.hg.1 | ENST00000604015.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014117.hg.1 | NR_026749.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100014117.hg.1 | NR_026749.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100014117.hg.1 | NR_026749.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100014117.hg.1 | NR_026749.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100014117.hg.1 | NR_026749.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100014117.hg.1 | NR_026749.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100014117.hg.1 | NR_026749.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100014117.hg.1 | NR_026749.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100014117.hg.1 | NR_026749.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100014117.hg.1 | NR_026749.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100014117.hg.1 | NR_026749.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100014117.hg.1 | NR_026749.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100014117.hg.1 | NR_026749.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100014117.hg.1 | NR_026749.2 | Refseq |

SKINT1L
Genomic Map