Assay Details
| Gene Symbol: TMCO4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017000918.1 | XP_016856407.1 | 3 - 4 | 585 | 65 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_017000918.1 transcripts.
Set Membership: |
|

TMCO4
Genomic Map