Assay Details
| Gene Symbol: SORT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006710812.1 | XP_006710875.1 | 1 - 2 | 129 | 102 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_006710812.1 transcripts.
Set Membership: |
|

SORT1
Genomic Map