Assay Details
| Gene Symbol: BROX | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001288579.1 | NP_001275508.1 | 12 | 3711 | 83 | |
| NM_001288580.1 | NP_001275509.1 | 12 | 3417 | 83 | ||
| NM_001288581.1 | NP_001275510.1 | 12 | 3353 | 83 | ||
| NM_144695.3 | NP_653296.2 | 13 | 3807 | 83 | ||
| XM_005273065.2 | XP_005273122.2 | 13 | 3443 | 83 | ||
| XM_005273069.4 | XP_005273126.1 | 12 | 3679 | 83 | ||
| XM_006711173.3 | XP_006711236.1 | 14 | 3876 | 83 | ||
| XM_011509212.2 | XP_011507514.1 | 12 | 3770 | 83 | ||
| XM_011509213.2 | XP_011507515.1 | 12 | 3751 | 83 | ||
| XM_011509214.2 | XP_011507516.1 | 14 | 3676 | 83 | ||
| XM_017000374.1 | XP_016855863.1 | 15 | 4001 | 83 | ||
| XM_017000375.1 | XP_016855864.1 | 11 | 3656 | 83 | ||
| XM_017000376.1 | XP_016855865.1 | 13 | 3804 | 83 | ||
| GenBank mRNA | AK025140.1 | - | 1 | 1603 | 83 | |
| AK098136.1 | - | 1 | 2020 | 83 | ||
More Information
Set Membership: |
|

BROX
Genomic Map