Assay Details
| Gene Symbol: SRSF10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001191005.2 | NP_001177934.1 | 6 | 4136 | 126 | |
| NM_054016.3 | NP_473357.1 | 6 | 4139 | 126 | ||
| GenBank mRNA | AK054635.1 | - | 1 | 1115 | 124 | |
| AL133655.1 | - | 1 | 474 | 125 | ||
More Information
Set Membership: |
|

SRSF10
Genomic Map