Assay Details
| Gene Symbol: LINC00862 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | EU250749.1 | - | 1 - 2 | 113 | 121 | |
More Information
Set Membership: |
|
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0100016885.hg.1 | ENST00000367355.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016885.hg.1 | ENST00000367355.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016885.hg.1 | ENST00000367355.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016885.hg.1 | ENST00000367355.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016885.hg.1 | ENST00000367355.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016885.hg.1 | ENST00000367355.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016885.hg.1 | ENST00000367355.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016885.hg.1 | ENST00000367355.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016885.hg.1 | ENST00000367355.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016885.hg.1 | ENST00000367355.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016885.hg.1 | ENST00000367355.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016885.hg.1 | ENST00000367355.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016885.hg.1 | ENST00000367355.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016885.hg.1 | ENST00000367355.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016885.hg.1 | ENST00000367355.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016885.hg.1 | ENST00000367355.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016885.hg.1 | ENST00000367356.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016885.hg.1 | ENST00000367356.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016885.hg.1 | ENST00000367356.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016885.hg.1 | ENST00000367356.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016885.hg.1 | ENST00000367356.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016885.hg.1 | ENST00000367356.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016885.hg.1 | NR_040064.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016885.hg.1 | NR_040064.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016885.hg.1 | NR_040064.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016885.hg.1 | NR_040064.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016885.hg.1 | NR_040064.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016885.hg.1 | NR_040064.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016885.hg.1 | NR_040064.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016885.hg.1 | NR_040064.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100016885.hg.1 | ENST00000367355.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016885.hg.1 | ENST00000367355.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016885.hg.1 | ENST00000367355.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016885.hg.1 | ENST00000367355.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016885.hg.1 | ENST00000367355.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016885.hg.1 | ENST00000367355.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016885.hg.1 | ENST00000367355.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016885.hg.1 | ENST00000367355.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016885.hg.1 | ENST00000367355.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016885.hg.1 | ENST00000367355.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016885.hg.1 | ENST00000367355.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016885.hg.1 | ENST00000367355.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016885.hg.1 | ENST00000367355.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016885.hg.1 | ENST00000367355.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016885.hg.1 | ENST00000367355.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016885.hg.1 | ENST00000367355.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016885.hg.1 | ENST00000367356.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016885.hg.1 | ENST00000367356.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016885.hg.1 | ENST00000367356.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016885.hg.1 | ENST00000367356.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016885.hg.1 | ENST00000367356.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016885.hg.1 | ENST00000367356.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016885.hg.1 | NR_040064.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100016885.hg.1 | NR_040064.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100016885.hg.1 | NR_040064.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100016885.hg.1 | NR_040064.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100016885.hg.1 | NR_040064.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100016885.hg.1 | NR_040064.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100016885.hg.1 | NR_040064.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100016885.hg.1 | NR_040064.1 | Refseq |

LINC00862
Genomic Map