Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017926.hg.1 |
ENST00000318160.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017926.hg.1 |
ENST00000318160.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017926.hg.1 |
ENST00000318160.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017926.hg.1 |
ENST00000318160.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017926.hg.1 |
ENST00000318160.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017926.hg.1 |
NM_022469.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017926.hg.1 |
NM_022469.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017926.hg.1 |
NM_022469.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017926.hg.1 |
NM_022469.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017926.hg.1 |
NM_022469.3 |
Refseq |