Assay Details
Gene Symbol: FCGR2C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK307162.1 | - | 1 - 2 | 173 | 81 | |
DA851200.1 | - | 1 - 2 | 208 | 81 | ||
DA859515.1 | - | 1 - 2 | 95 | 81 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC0100018309.hg.1 | ENST00000465075.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100018309.hg.1 | ENST00000465075.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100018309.hg.1 | ENST00000465075.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100018309.hg.1 | ENST00000465075.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100018309.hg.1 | ENST00000465075.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100018309.hg.1 | ENST00000465075.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100018309.hg.1 | ENST00000465075.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100018309.hg.1 | ENST00000466542.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100018309.hg.1 | ENST00000466542.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100018309.hg.1 | ENST00000466542.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100018309.hg.1 | ENST00000466542.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100018309.hg.1 | ENST00000466542.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100018309.hg.1 | ENST00000466542.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100018309.hg.1 | ENST00000466542.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100018309.hg.1 | ENST00000466542.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100018309.hg.1 | ENST00000466542.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100018309.hg.1 | ENST00000466542.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100018309.hg.1 | ENST00000467903.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100018309.hg.1 | ENST00000467903.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100018309.hg.1 | ENST00000467903.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100018309.hg.1 | ENST00000467903.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100018309.hg.1 | ENST00000467903.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100018309.hg.1 | ENST00000467903.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100018309.hg.1 | ENST00000473530.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100018309.hg.1 | ENST00000473530.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100018309.hg.1 | ENST00000473530.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100018309.hg.1 | ENST00000473530.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100018309.hg.1 | ENST00000473530.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100018309.hg.1 | ENST00000473530.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100018309.hg.1 | ENST00000473530.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100018309.hg.1 | ENST00000473530.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100018309.hg.1 | ENST00000473530.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100018309.hg.1 | ENST00000473530.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100018309.hg.1 | ENST00000473530.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100018309.hg.1 | ENST00000473712.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100018309.hg.1 | ENST00000473712.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100018309.hg.1 | ENST00000473712.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100018309.hg.1 | ENST00000473712.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100018309.hg.1 | ENST00000473712.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100018309.hg.1 | ENST00000482226.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100018309.hg.1 | ENST00000482226.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100018309.hg.1 | ENST00000482226.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100018309.hg.1 | ENST00000482226.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100018309.hg.1 | ENST00000496692.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100018309.hg.1 | ENST00000496692.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100018309.hg.1 | ENST00000496692.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100018309.hg.1 | ENST00000496692.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100018309.hg.1 | ENST00000496692.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100018309.hg.1 | ENST00000496692.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100018309.hg.1 | ENST00000502411.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100018309.hg.1 | ENST00000502411.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100018309.hg.1 | ENST00000502411.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100018309.hg.1 | ENST00000502411.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100018309.hg.1 | ENST00000502411.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100018309.hg.1 | ENST00000502411.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100018309.hg.1 | ENST00000507374.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100018309.hg.1 | ENST00000507374.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100018309.hg.1 | ENST00000507374.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100018309.hg.1 | ENST00000507374.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100018309.hg.1 | ENST00000507374.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100018309.hg.1 | ENST00000508651.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100018309.hg.1 | ENST00000508651.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100018309.hg.1 | ENST00000508651.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100018309.hg.1 | ENST00000508651.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100018309.hg.1 | ENST00000508651.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100018309.hg.1 | ENST00000543859.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100018309.hg.1 | ENST00000543859.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100018309.hg.1 | ENST00000543859.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100018309.hg.1 | ENST00000543859.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100018309.hg.1 | ENST00000543859.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100018309.hg.1 | ENST00000543859.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100018309.hg.1 | ENST00000543859.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100018309.hg.1 | ENST00000543859.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100018309.hg.1 | ENST00000543859.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100018309.hg.1 | ENST00000543859.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100018309.hg.1 | ENST00000543859.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100018309.hg.1 | ENST00000543859.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100018309.hg.1 | ENST00000611236.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100018309.hg.1 | ENST00000611236.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100018309.hg.1 | ENST00000611236.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100018309.hg.1 | ENST00000611236.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100018309.hg.1 | ENST00000611236.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100018309.hg.1 | ENST00000611236.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100018309.hg.1 | ENST00000611236.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100018309.hg.1 | ENST00000611236.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100018309.hg.1 | NR_047648.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100018309.hg.1 | NR_047648.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100018309.hg.1 | NR_047648.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100018309.hg.1 | NR_047648.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100018309.hg.1 | NR_047648.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100018309.hg.1 | NR_047648.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100018309.hg.1 | NR_047648.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100018309.hg.1 | NR_047648.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100018309.hg.1 | NR_047648.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100018309.hg.1 | NR_047648.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100018309.hg.1 | NR_047648.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100018309.hg.1 | NR_047648.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100018309.hg.1 | NR_047648.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100018309.hg.1 | ENST00000465075.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100018309.hg.1 | ENST00000465075.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100018309.hg.1 | ENST00000465075.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100018309.hg.1 | ENST00000465075.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100018309.hg.1 | ENST00000465075.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100018309.hg.1 | ENST00000465075.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100018309.hg.1 | ENST00000465075.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100018309.hg.1 | ENST00000466542.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100018309.hg.1 | ENST00000466542.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100018309.hg.1 | ENST00000466542.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100018309.hg.1 | ENST00000466542.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100018309.hg.1 | ENST00000466542.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100018309.hg.1 | ENST00000466542.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100018309.hg.1 | ENST00000466542.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100018309.hg.1 | ENST00000466542.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100018309.hg.1 | ENST00000466542.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100018309.hg.1 | ENST00000466542.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100018309.hg.1 | ENST00000467903.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100018309.hg.1 | ENST00000467903.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100018309.hg.1 | ENST00000467903.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100018309.hg.1 | ENST00000467903.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100018309.hg.1 | ENST00000467903.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100018309.hg.1 | ENST00000467903.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100018309.hg.1 | ENST00000473530.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100018309.hg.1 | ENST00000473530.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100018309.hg.1 | ENST00000473530.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100018309.hg.1 | ENST00000473530.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100018309.hg.1 | ENST00000473530.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100018309.hg.1 | ENST00000473530.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100018309.hg.1 | ENST00000473530.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100018309.hg.1 | ENST00000473530.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100018309.hg.1 | ENST00000473530.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100018309.hg.1 | ENST00000473530.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100018309.hg.1 | ENST00000473530.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100018309.hg.1 | ENST00000473712.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100018309.hg.1 | ENST00000473712.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100018309.hg.1 | ENST00000473712.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100018309.hg.1 | ENST00000473712.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100018309.hg.1 | ENST00000473712.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100018309.hg.1 | ENST00000482226.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100018309.hg.1 | ENST00000482226.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100018309.hg.1 | ENST00000482226.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100018309.hg.1 | ENST00000482226.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100018309.hg.1 | ENST00000496692.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100018309.hg.1 | ENST00000496692.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100018309.hg.1 | ENST00000496692.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100018309.hg.1 | ENST00000496692.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100018309.hg.1 | ENST00000496692.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100018309.hg.1 | ENST00000496692.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100018309.hg.1 | ENST00000502411.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100018309.hg.1 | ENST00000502411.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100018309.hg.1 | ENST00000502411.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100018309.hg.1 | ENST00000502411.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100018309.hg.1 | ENST00000502411.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100018309.hg.1 | ENST00000502411.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100018309.hg.1 | ENST00000507374.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100018309.hg.1 | ENST00000507374.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100018309.hg.1 | ENST00000507374.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100018309.hg.1 | ENST00000507374.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100018309.hg.1 | ENST00000507374.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100018309.hg.1 | ENST00000508651.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100018309.hg.1 | ENST00000508651.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100018309.hg.1 | ENST00000508651.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100018309.hg.1 | ENST00000508651.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100018309.hg.1 | ENST00000508651.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100018309.hg.1 | ENST00000543859.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100018309.hg.1 | ENST00000543859.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100018309.hg.1 | ENST00000543859.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100018309.hg.1 | ENST00000543859.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100018309.hg.1 | ENST00000543859.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100018309.hg.1 | ENST00000543859.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100018309.hg.1 | ENST00000543859.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100018309.hg.1 | ENST00000543859.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100018309.hg.1 | ENST00000543859.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100018309.hg.1 | ENST00000543859.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100018309.hg.1 | ENST00000543859.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100018309.hg.1 | ENST00000543859.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100018309.hg.1 | ENST00000611236.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100018309.hg.1 | ENST00000611236.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100018309.hg.1 | ENST00000611236.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100018309.hg.1 | ENST00000611236.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100018309.hg.1 | ENST00000611236.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100018309.hg.1 | ENST00000611236.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100018309.hg.1 | ENST00000611236.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100018309.hg.1 | ENST00000611236.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100018309.hg.1 | NR_047648.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100018309.hg.1 | NR_047648.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100018309.hg.1 | NR_047648.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100018309.hg.1 | NR_047648.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100018309.hg.1 | NR_047648.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100018309.hg.1 | NR_047648.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100018309.hg.1 | NR_047648.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100018309.hg.1 | NR_047648.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100018309.hg.1 | NR_047648.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100018309.hg.1 | NR_047648.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100018309.hg.1 | NR_047648.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100018309.hg.1 | NR_047648.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100018309.hg.1 | NR_047648.1 | Refseq |
