Assay Details
| Gene Symbol: LOC79160 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_125359.1 | 1 | 722 | 112 | ||
| GenBank mRNA | BC002831.2 | - | 1 | 722 | 3636069 | 112 |
| BU624279.1 | - | 1 | 112 | |||
More Information
Set Membership: |
|

LOC79160
Genomic Map