Assay Details
| Gene Symbol: GDF5OS | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | BC085019.1 | - | 1 - 2 | 139 | 6650156 | 101 |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC085019.1 transcripts.
Set Membership: |
|

GDF5OS
Genomic Map