Assay Details
| Gene Symbol: ZNF341-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AW136575.1 | - | 1 - 2 | 427 | 79 | |
More Information
Set Membership: |
|
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC2000008899.hg.1 | ENST00000423074.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008899.hg.1 | ENST00000423074.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008899.hg.1 | ENST00000423074.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008899.hg.1 | ENST00000423074.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008899.hg.1 | ENST00000423074.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008899.hg.1 | ENST00000439444.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008899.hg.1 | ENST00000439444.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008899.hg.1 | ENST00000439444.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008899.hg.1 | ENST00000439444.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008899.hg.1 | ENST00000439444.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008899.hg.1 | ENST00000443171.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008899.hg.1 | ENST00000443171.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008899.hg.1 | ENST00000443171.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008899.hg.1 | ENST00000443171.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008899.hg.1 | ENST00000443171.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008899.hg.1 | ENST00000443171.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008899.hg.1 | ENST00000443171.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008899.hg.1 | ENST00000443171.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008899.hg.1 | ENST00000443171.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008899.hg.1 | ENST00000443171.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008899.hg.1 | ENST00000443171.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008899.hg.1 | ENST00000443171.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008899.hg.1 | ENST00000443171.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008899.hg.1 | ENST00000443171.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008899.hg.1 | NR_110623.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008899.hg.1 | NR_110623.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008899.hg.1 | NR_110623.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008899.hg.1 | NR_110623.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008899.hg.1 | NR_110623.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008899.hg.1 | NR_110623.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008899.hg.1 | NR_110623.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000008899.hg.1 | ENST00000423074.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000008899.hg.1 | ENST00000423074.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000008899.hg.1 | ENST00000423074.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000008899.hg.1 | ENST00000423074.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000008899.hg.1 | ENST00000423074.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000008899.hg.1 | ENST00000439444.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000008899.hg.1 | ENST00000439444.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000008899.hg.1 | ENST00000439444.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000008899.hg.1 | ENST00000439444.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000008899.hg.1 | ENST00000439444.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000008899.hg.1 | ENST00000443171.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000008899.hg.1 | ENST00000443171.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000008899.hg.1 | ENST00000443171.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000008899.hg.1 | ENST00000443171.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000008899.hg.1 | ENST00000443171.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000008899.hg.1 | ENST00000443171.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000008899.hg.1 | ENST00000443171.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000008899.hg.1 | ENST00000443171.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000008899.hg.1 | ENST00000443171.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000008899.hg.1 | ENST00000443171.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000008899.hg.1 | ENST00000443171.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000008899.hg.1 | ENST00000443171.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000008899.hg.1 | ENST00000443171.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000008899.hg.1 | ENST00000443171.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000008899.hg.1 | NR_110623.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000008899.hg.1 | NR_110623.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000008899.hg.1 | NR_110623.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000008899.hg.1 | NR_110623.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000008899.hg.1 | NR_110623.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000008899.hg.1 | NR_110623.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000008899.hg.1 | NR_110623.1 | Refseq |

ZNF341-AS1
Genomic Map