Assay Details
| Gene Symbol: SAMHD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_015474.3 | NP_056289.2 | 12 - 13 | 1609 | 95 | |
| XM_005260384.3 | XP_005260441.1 | 12 - 13 | 1582 | 95 | ||
| XM_011528761.2 | XP_011527063.1 | 12 - 13 | 1582 | 95 | ||
| GenBank mRNA | AB013847.1 | - | 12 - 13 | 1409 | 95 | |
| AB208944.1 | - | 12 - 13 | 1434 | 95 | ||
| AF228421.1 | - | 12 - 13 | 1562 | 95 | ||
| AK027811.1 | - | 12 - 13 | 1582 | 95 | ||
| AK304187.1 | - | 12 - 13 | 1366 | 95 | ||
| AK311150.1 | - | 1 - 2 | 44 | 95 | ||
| AK315169.1 | - | 12 - 13 | 1542 | 95 | ||
| AL050267.1 | - | 12 - 13 | 1484 | 95 | ||
| BC036450.2 | - | 12 - 13 | 1437 | 4816438 | 95 | |
More Information
Set Membership: |
|

SAMHD1
Genomic Map