Assay Details
| Gene Symbol: HSD17B7P2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AF165514.1 | - | 6 - 7 | 767 | 98 | |
| AF165517.1 | - | 5 - 6 | 717 | 98 | ||
| AK293968.1 | - | 5 - 6 | 714 | 98 | ||
More Information
Set Membership: |
|
Panther Classification:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0100010389.hg.1 | ENST00000254521.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010389.hg.1 | ENST00000254521.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010389.hg.1 | ENST00000254521.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010389.hg.1 | ENST00000254521.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010389.hg.1 | ENST00000254521.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010389.hg.1 | ENST00000254521.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010389.hg.1 | ENST00000254521.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010389.hg.1 | ENST00000254521.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010389.hg.1 | ENST00000254521.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010389.hg.1 | ENST00000254521.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010389.hg.1 | ENST00000254521.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010389.hg.1 | ENST00000254521.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010389.hg.1 | ENST00000254521.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010389.hg.1 | ENST00000367913.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010389.hg.1 | ENST00000367913.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010389.hg.1 | ENST00000367913.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010389.hg.1 | ENST00000367913.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010389.hg.1 | ENST00000367913.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010389.hg.1 | ENST00000367915.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010389.hg.1 | ENST00000367915.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010389.hg.1 | ENST00000367915.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010389.hg.1 | ENST00000367915.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010389.hg.1 | ENST00000463037.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010389.hg.1 | ENST00000463037.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010389.hg.1 | ENST00000463037.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010389.hg.1 | ENST00000463037.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010389.hg.1 | ENST00000463037.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010389.hg.1 | ENST00000463037.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010389.hg.1 | ENST00000463037.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010389.hg.1 | ENST00000463037.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010389.hg.1 | ENST00000463037.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010389.hg.1 | ENST00000463037.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010389.hg.1 | ENST00000466176.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010389.hg.1 | ENST00000466176.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010389.hg.1 | ENST00000466176.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010389.hg.1 | ENST00000466176.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010389.hg.1 | ENST00000466176.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010389.hg.1 | ENST00000466176.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010389.hg.1 | ENST00000466176.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010389.hg.1 | ENST00000466176.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010389.hg.1 | ENST00000470195.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010389.hg.1 | ENST00000470195.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010389.hg.1 | ENST00000470195.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010389.hg.1 | ENST00000470195.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010389.hg.1 | ENST00000470195.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010389.hg.1 | ENST00000470195.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010389.hg.1 | ENST00000484251.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010389.hg.1 | ENST00000484251.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010389.hg.1 | ENST00000484251.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010389.hg.1 | ENST00000484251.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010389.hg.1 | ENST00000484251.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010389.hg.1 | ENST00000485405.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010389.hg.1 | ENST00000485405.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010389.hg.1 | ENST00000485405.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010389.hg.1 | ENST00000485405.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010389.hg.1 | ENST00000485405.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010389.hg.1 | ENST00000485405.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010389.hg.1 | ENST00000485405.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010389.hg.1 | ENST00000485405.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010389.hg.1 | ENST00000485405.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010389.hg.1 | ENST00000485405.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010389.hg.1 | ENST00000485405.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010389.hg.1 | ENST00000485405.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010389.hg.1 | ENST00000485405.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010389.hg.1 | ENST00000485405.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010389.hg.1 | ENST00000485405.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010389.hg.1 | ENST00000485405.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010389.hg.1 | ENST00000488656.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010389.hg.1 | ENST00000488656.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010389.hg.1 | ENST00000488656.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010389.hg.1 | ENST00000494450.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010389.hg.1 | ENST00000494450.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010389.hg.1 | ENST00000494450.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010389.hg.1 | ENST00000494450.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010389.hg.1 | ENST00000494450.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010389.hg.1 | NM_001304512.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010389.hg.1 | NM_001304512.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010389.hg.1 | NM_001304512.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010389.hg.1 | NM_001304512.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010389.hg.1 | NM_001304512.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010389.hg.1 | NM_001304512.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010389.hg.1 | NM_001304512.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010389.hg.1 | NM_001304513.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010389.hg.1 | NM_001304513.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010389.hg.1 | NM_001304513.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010389.hg.1 | NM_001304513.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010389.hg.1 | NM_001304513.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010389.hg.1 | NM_001304513.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010389.hg.1 | NM_016371.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010389.hg.1 | NM_016371.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010389.hg.1 | NM_016371.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010389.hg.1 | NM_016371.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010389.hg.1 | NM_016371.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010389.hg.1 | NM_016371.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010389.hg.1 | NM_016371.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010389.hg.1 | NM_016371.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010389.hg.1 | NM_016371.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010389.hg.1 | NM_016371.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010389.hg.1 | NM_016371.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010389.hg.1 | NM_016371.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010389.hg.1 | NM_016371.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010389.hg.1 | NM_016371.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007346.hg.1 | ENST00000471365.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007346.hg.1 | ENST00000471365.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007346.hg.1 | ENST00000471365.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007346.hg.1 | ENST00000471365.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007346.hg.1 | ENST00000471365.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007346.hg.1 | ENST00000471365.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007346.hg.1 | ENST00000471365.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007346.hg.1 | ENST00000471365.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007346.hg.1 | ENST00000471365.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007346.hg.1 | ENST00000494540.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007346.hg.1 | ENST00000494540.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007346.hg.1 | ENST00000494540.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007346.hg.1 | ENST00000494540.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007346.hg.1 | ENST00000494540.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007346.hg.1 | ENST00000494540.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007346.hg.1 | ENST00000494540.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007346.hg.1 | ENST00000494540.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007346.hg.1 | ENST00000494540.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007346.hg.1 | ENST00000494540.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007346.hg.1 | NR_003086.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007346.hg.1 | NR_003086.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007346.hg.1 | NR_003086.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007346.hg.1 | NR_003086.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007346.hg.1 | NR_003086.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007346.hg.1 | NR_003086.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007346.hg.1 | NR_003086.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007346.hg.1 | NR_003086.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007346.hg.1 | NR_003086.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007346.hg.1 | NR_003086.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000007346.hg.1 | ENST00000471365.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007346.hg.1 | ENST00000471365.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007346.hg.1 | ENST00000471365.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007346.hg.1 | ENST00000471365.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007346.hg.1 | ENST00000471365.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007346.hg.1 | ENST00000471365.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007346.hg.1 | ENST00000471365.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007346.hg.1 | ENST00000471365.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007346.hg.1 | ENST00000471365.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007346.hg.1 | ENST00000494540.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007346.hg.1 | ENST00000494540.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007346.hg.1 | ENST00000494540.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007346.hg.1 | ENST00000494540.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007346.hg.1 | ENST00000494540.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007346.hg.1 | ENST00000494540.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007346.hg.1 | ENST00000494540.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007346.hg.1 | ENST00000494540.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007346.hg.1 | ENST00000494540.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007346.hg.1 | ENST00000494540.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007346.hg.1 | NR_003086.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000007346.hg.1 | NR_003086.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000007346.hg.1 | NR_003086.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000007346.hg.1 | NR_003086.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000007346.hg.1 | NR_003086.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000007346.hg.1 | NR_003086.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000007346.hg.1 | NR_003086.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000007346.hg.1 | NR_003086.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000007346.hg.1 | NR_003086.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000007346.hg.1 | NR_003086.1 | Refseq |

HSD17B7P2
Genomic Map