| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009937.hg.1 |
ENST00000411453.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009937.hg.1 |
ENST00000411453.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009937.hg.1 |
ENST00000411453.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009937.hg.1 |
ENST00000411453.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009937.hg.1 |
ENST00000411453.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009937.hg.1 |
ENST00000411453.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009937.hg.1 |
ENST00000411453.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009937.hg.1 |
ENST00000427333.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009937.hg.1 |
ENST00000427333.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009937.hg.1 |
ENST00000427333.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009937.hg.1 |
ENST00000427333.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009937.hg.1 |
ENST00000427333.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009937.hg.1 |
ENST00000427333.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009937.hg.1 |
NR_109943.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009937.hg.1 |
NR_109943.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009937.hg.1 |
NR_109943.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009937.hg.1 |
NR_109943.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000009937.hg.1 |
ENST00000411453.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000009937.hg.1 |
ENST00000411453.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000009937.hg.1 |
ENST00000411453.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000009937.hg.1 |
ENST00000411453.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000009937.hg.1 |
ENST00000411453.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000009937.hg.1 |
ENST00000411453.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000009937.hg.1 |
ENST00000411453.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000009937.hg.1 |
ENST00000427333.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000009937.hg.1 |
ENST00000427333.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000009937.hg.1 |
ENST00000427333.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000009937.hg.1 |
ENST00000427333.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000009937.hg.1 |
ENST00000427333.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000009937.hg.1 |
ENST00000427333.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000009937.hg.1 |
NR_109943.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000009937.hg.1 |
NR_109943.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000009937.hg.1 |
NR_109943.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000009937.hg.1 |
NR_109943.1 |
Refseq |