Assay Details
Gene Symbol: DSCR4-IT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | BG462133.1 | - | 333 | 91 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
MicroArray Probes:
