Assay Details
| Gene Symbol: LOC284825 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_126010.1 | 1 - 2 | 126 | 99 | ||
| GenBank mRNA | AK093119.1 | - | 1 - 2 | 126 | 99 | |
| BC039446.1 | - | 1 - 2 | 93 | 99 | ||
More Information
Set Membership: |
|
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC2100006834.hg.1 | ENST00000423808.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006834.hg.1 | ENST00000423808.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006834.hg.1 | ENST00000423808.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006834.hg.1 | ENST00000423808.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006834.hg.1 | ENST00000423808.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006834.hg.1 | ENST00000423808.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006834.hg.1 | ENST00000423808.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006834.hg.1 | ENST00000423808.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006834.hg.1 | ENST00000423808.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006834.hg.1 | ENST00000423808.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006834.hg.1 | ENST00000423808.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006834.hg.1 | ENST00000423808.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006834.hg.1 | ENST00000423808.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006834.hg.1 | ENST00000423808.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006834.hg.1 | ENST00000423808.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006834.hg.1 | ENST00000426534.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006834.hg.1 | ENST00000426534.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006834.hg.1 | ENST00000426534.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006834.hg.1 | ENST00000426534.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006834.hg.1 | ENST00000426534.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006834.hg.1 | ENST00000426534.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006834.hg.1 | ENST00000426534.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006834.hg.1 | ENST00000426534.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006834.hg.1 | ENST00000426534.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006834.hg.1 | ENST00000426534.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006834.hg.1 | ENST00000426534.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006834.hg.1 | ENST00000426534.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006834.hg.1 | ENST00000426534.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006834.hg.1 | ENST00000426534.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006834.hg.1 | ENST00000426534.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006834.hg.1 | ENST00000426534.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006834.hg.1 | ENST00000426534.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006834.hg.1 | ENST00000426534.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006834.hg.1 | ENST00000426534.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006834.hg.1 | ENST00000426534.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006834.hg.1 | ENST00000436878.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006834.hg.1 | ENST00000436878.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006834.hg.1 | ENST00000436878.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006834.hg.1 | ENST00000436878.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006834.hg.1 | ENST00000436878.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006834.hg.1 | ENST00000436878.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006834.hg.1 | ENST00000436878.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006834.hg.1 | ENST00000458316.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006834.hg.1 | ENST00000458316.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006834.hg.1 | ENST00000458316.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006834.hg.1 | ENST00000458316.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006834.hg.1 | ENST00000458316.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006834.hg.1 | ENST00000458316.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006834.hg.1 | ENST00000458316.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006834.hg.1 | ENST00000609782.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006834.hg.1 | ENST00000609782.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006834.hg.1 | ENST00000609782.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006834.hg.1 | ENST00000609782.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006834.hg.1 | ENST00000616647.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006834.hg.1 | ENST00000616647.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006834.hg.1 | ENST00000616647.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006834.hg.1 | ENST00000616647.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006834.hg.1 | ENST00000616647.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006834.hg.1 | ENST00000616647.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006834.hg.1 | ENST00000616647.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006834.hg.1 | ENST00000631186.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006834.hg.1 | ENST00000631186.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006834.hg.1 | ENST00000631186.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006834.hg.1 | ENST00000631186.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006834.hg.1 | NR_126010.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006834.hg.1 | NR_126010.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006834.hg.1 | NR_126010.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006834.hg.1 | NR_126010.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006834.hg.1 | NR_126010.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006834.hg.1 | NR_126010.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006834.hg.1 | NR_126010.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006834.hg.1 | NR_126010.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006834.hg.1 | NR_126010.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006834.hg.1 | NR_126010.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006834.hg.1 | NR_126011.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006834.hg.1 | NR_126011.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006834.hg.1 | NR_126011.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006834.hg.1 | NR_126011.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006834.hg.1 | NR_126011.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006834.hg.1 | NR_126011.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006834.hg.1 | NR_126011.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006834.hg.1 | NR_126011.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012759.hg.1 | ENST00000488190.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012759.hg.1 | ENST00000488190.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012759.hg.1 | ENST00000488190.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012759.hg.1 | ENST00000488190.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012759.hg.1 | ENST00000488190.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012759.hg.1 | ENST00000488190.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012759.hg.1 | ENST00000488190.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012759.hg.1 | ENST00000488190.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012759.hg.1 | ENST00000488190.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012759.hg.1 | ENST00000488190.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006834.hg.1 | ENST00000423808.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006834.hg.1 | ENST00000423808.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006834.hg.1 | ENST00000423808.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006834.hg.1 | ENST00000423808.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006834.hg.1 | ENST00000423808.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006834.hg.1 | ENST00000423808.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006834.hg.1 | ENST00000423808.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006834.hg.1 | ENST00000423808.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006834.hg.1 | ENST00000423808.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006834.hg.1 | ENST00000423808.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006834.hg.1 | ENST00000423808.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006834.hg.1 | ENST00000423808.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006834.hg.1 | ENST00000423808.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006834.hg.1 | ENST00000423808.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006834.hg.1 | ENST00000423808.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006834.hg.1 | ENST00000426534.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006834.hg.1 | ENST00000426534.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006834.hg.1 | ENST00000426534.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006834.hg.1 | ENST00000426534.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006834.hg.1 | ENST00000426534.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006834.hg.1 | ENST00000426534.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006834.hg.1 | ENST00000426534.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006834.hg.1 | ENST00000426534.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006834.hg.1 | ENST00000426534.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006834.hg.1 | ENST00000426534.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006834.hg.1 | ENST00000426534.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006834.hg.1 | ENST00000426534.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006834.hg.1 | ENST00000426534.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006834.hg.1 | ENST00000426534.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006834.hg.1 | ENST00000426534.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006834.hg.1 | ENST00000426534.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006834.hg.1 | ENST00000426534.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006834.hg.1 | ENST00000426534.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006834.hg.1 | ENST00000426534.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006834.hg.1 | ENST00000426534.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006834.hg.1 | ENST00000436878.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006834.hg.1 | ENST00000436878.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006834.hg.1 | ENST00000436878.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006834.hg.1 | ENST00000436878.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006834.hg.1 | ENST00000436878.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006834.hg.1 | ENST00000436878.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006834.hg.1 | ENST00000436878.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006834.hg.1 | ENST00000458316.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006834.hg.1 | ENST00000458316.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006834.hg.1 | ENST00000458316.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006834.hg.1 | ENST00000458316.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006834.hg.1 | ENST00000458316.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006834.hg.1 | ENST00000458316.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006834.hg.1 | ENST00000458316.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006834.hg.1 | ENST00000609782.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006834.hg.1 | ENST00000609782.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006834.hg.1 | ENST00000609782.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006834.hg.1 | ENST00000609782.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006834.hg.1 | ENST00000616647.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006834.hg.1 | ENST00000616647.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006834.hg.1 | ENST00000616647.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006834.hg.1 | ENST00000616647.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006834.hg.1 | ENST00000616647.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006834.hg.1 | ENST00000616647.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006834.hg.1 | ENST00000616647.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006834.hg.1 | ENST00000631186.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006834.hg.1 | ENST00000631186.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006834.hg.1 | ENST00000631186.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006834.hg.1 | ENST00000631186.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006834.hg.1 | NR_126010.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100006834.hg.1 | NR_126010.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100006834.hg.1 | NR_126010.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100006834.hg.1 | NR_126010.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100006834.hg.1 | NR_126010.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100006834.hg.1 | NR_126010.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100006834.hg.1 | NR_126010.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100006834.hg.1 | NR_126010.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100006834.hg.1 | NR_126010.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100006834.hg.1 | NR_126010.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100006834.hg.1 | NR_126011.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100006834.hg.1 | NR_126011.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100006834.hg.1 | NR_126011.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100006834.hg.1 | NR_126011.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100006834.hg.1 | NR_126011.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100006834.hg.1 | NR_126011.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100006834.hg.1 | NR_126011.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100006834.hg.1 | NR_126011.1 | Refseq |

LOC284825
Genomic Map