Assay Details
| Gene Symbol: MDK | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001270552.1 | NP_001257481.1 | 1 - 2 | 99 | 127 | |
| NM_002391.4 | NP_002382.1 | 1 - 2 | 99 | 127 | ||
| GenBank mRNA | BM761575.1 | - | 1 - 2 | 85 | 127 | |
| BM887813.1 | - | 1 - 2 | 99 | 127 | ||
| DQ323888.1 | - | 1 - 2 | 85 | 127 | ||
| DQ323889.1 | - | 1 - 2 | 85 | 127 | ||
| DQ323890.1 | - | 1 - 2 | 85 | 127 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the DQ323888.1, DQ323889.1, DQ323890.1, NM_001270552.1, NM_002391.4 transcripts.
Set Membership: |
|

MDK
Genomic Map