| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100008037.hg.1 |
ENST00000335987.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100008037.hg.1 |
ENST00000335987.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100008037.hg.1 |
ENST00000335987.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100008037.hg.1 |
ENST00000335987.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100008037.hg.1 |
ENST00000335987.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100008037.hg.1 |
ENST00000335987.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100008037.hg.1 |
ENST00000335987.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100008037.hg.1 |
ENST00000335987.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100008037.hg.1 |
ENST00000531907.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100008037.hg.1 |
ENST00000531907.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100008037.hg.1 |
ENST00000531907.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100008037.hg.1 |
ENST00000532448.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100008037.hg.1 |
ENST00000532448.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100008037.hg.1 |
ENST00000532448.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100008037.hg.1 |
ENST00000532448.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100008037.hg.1 |
ENST00000532448.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100008037.hg.1 |
ENST00000532448.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100008037.hg.1 |
NM_004561.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100008037.hg.1 |
NM_004561.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100008037.hg.1 |
NM_004561.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100008037.hg.1 |
NM_004561.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100008037.hg.1 |
NM_004561.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100008037.hg.1 |
NM_004561.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100008037.hg.1 |
NM_004561.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100008037.hg.1 |
NM_004561.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100008037.hg.1 |
ENST00000335987.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100008037.hg.1 |
ENST00000335987.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100008037.hg.1 |
ENST00000335987.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100008037.hg.1 |
ENST00000335987.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100008037.hg.1 |
ENST00000335987.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100008037.hg.1 |
ENST00000335987.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100008037.hg.1 |
ENST00000335987.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100008037.hg.1 |
ENST00000335987.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100008037.hg.1 |
ENST00000531907.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100008037.hg.1 |
ENST00000531907.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100008037.hg.1 |
ENST00000531907.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100008037.hg.1 |
ENST00000532448.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100008037.hg.1 |
ENST00000532448.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100008037.hg.1 |
ENST00000532448.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100008037.hg.1 |
ENST00000532448.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100008037.hg.1 |
ENST00000532448.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100008037.hg.1 |
ENST00000532448.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100008037.hg.1 |
NM_004561.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100008037.hg.1 |
NM_004561.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100008037.hg.1 |
NM_004561.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100008037.hg.1 |
NM_004561.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100008037.hg.1 |
NM_004561.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100008037.hg.1 |
NM_004561.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100008037.hg.1 |
NM_004561.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100008037.hg.1 |
NM_004561.3 |
Refseq |