Assay Details
| Gene Symbol: CREB3L1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_052854.3 | NP_443086.1 | 10 - 11 | 1729 | 89 | |
| GenBank mRNA | AB063321.1 | - | 10 - 11 | 1350 | 89 | |
| AF055009.1 | - | 2 - 3 | 849 | 89 | ||
| AK075538.1 | - | 5 - 6 | 500 | 89 | ||
| AK314072.1 | - | 10 - 11 | 1707 | 89 | ||
| BC014097.1 | - | 10 - 11 | 1372 | 4546950 | 89 | |
| BC015781.1 | - | 10 - 11 | 1358 | 4856946 | 89 | |
More Information
Set Membership: |
|

CREB3L1
Genomic Map