Assay Details
| Gene Symbol: TSKU | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001258210.1 | NP_001245139.1 | 2 | 947 | 157 | |
| NM_001318477.1 | NP_001305406.1 | 2 | 1844 | 157 | ||
| NM_001318478.1 | NP_001305407.1 | 2 | 1044 | 157 | ||
| NM_001318479.1 | NP_001305408.1 | 2 | 855 | 157 | ||
| NM_015516.3 | NP_056331.2 | 2 | 990 | 157 | ||
| GenBank mRNA | AF191019.1 | - | 1 | 831 | 157 | |
| AK075476.1 | - | 2 | 902 | 157 | ||
| AK091475.1 | - | 3 | 807 | 157 | ||
| AK126536.1 | - | 2 | 1844 | 157 | ||
| AK313405.1 | - | 2 | 903 | 157 | ||
| AL110276.1 | - | 1 | 486 | 157 | ||
| AY358317.1 | - | 2 | 881 | 157 | ||
| BC020975.1 | - | 2 | 868 | 3848274 | 157 | |
| BT007440.1 | - | 1 | 818 | 157 | ||
More Information
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1100008514.hg.1 | ENST00000333090.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008514.hg.1 | ENST00000333090.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008514.hg.1 | ENST00000333090.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008514.hg.1 | ENST00000333090.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008514.hg.1 | ENST00000333090.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008514.hg.1 | ENST00000333090.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008514.hg.1 | ENST00000525167.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008514.hg.1 | ENST00000525167.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008514.hg.1 | ENST00000527881.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008514.hg.1 | ENST00000527881.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008514.hg.1 | ENST00000527881.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008514.hg.1 | ENST00000527881.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008514.hg.1 | ENST00000527881.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008514.hg.1 | ENST00000533752.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008514.hg.1 | ENST00000533752.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008514.hg.1 | ENST00000533752.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008514.hg.1 | ENST00000533752.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008514.hg.1 | ENST00000612930.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008514.hg.1 | ENST00000612930.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008514.hg.1 | ENST00000612930.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008514.hg.1 | ENST00000612930.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008514.hg.1 | ENST00000612930.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008514.hg.1 | NM_001258210.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008514.hg.1 | NM_001258210.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008514.hg.1 | NM_001258210.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008514.hg.1 | NM_001258210.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008514.hg.1 | NM_001258210.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008514.hg.1 | NM_015516.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008514.hg.1 | NM_015516.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008514.hg.1 | NM_015516.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008514.hg.1 | NM_015516.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008514.hg.1 | NM_015516.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008514.hg.1 | NM_015516.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200006721.hg.1 | ENST00000417473.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200006721.hg.1 | ENST00000417473.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200006721.hg.1 | ENST00000417473.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200006721.hg.1 | ENST00000417473.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200006721.hg.1 | ENST00000417473.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200006721.hg.1 | ENST00000417473.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200006721.hg.1 | ENST00000417473.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200006721.hg.1 | ENST00000417473.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200006721.hg.1 | ENST00000417473.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200006721.hg.1 | ENST00000417473.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200006721.hg.1 | ENST00000417473.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200006721.hg.1 | ENST00000417473.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200006721.hg.1 | ENST00000417473.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200006721.hg.1 | ENST00000417473.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200006721.hg.1 | ENST00000417473.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200006721.hg.1 | ENST00000417473.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200006721.hg.1 | ENST00000432665.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200006721.hg.1 | ENST00000432665.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200006721.hg.1 | ENST00000432665.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200006721.hg.1 | ENST00000432665.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200006721.hg.1 | ENST00000432665.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200006721.hg.1 | ENST00000437795.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200006721.hg.1 | ENST00000437795.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200006721.hg.1 | ENST00000437795.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200006721.hg.1 | ENST00000437795.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200006721.hg.1 | ENST00000437795.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200006721.hg.1 | ENST00000437795.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200006721.hg.1 | ENST00000437795.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200006721.hg.1 | ENST00000437795.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200006721.hg.1 | ENST00000437795.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200006721.hg.1 | ENST00000437795.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200006721.hg.1 | ENST00000437795.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200006721.hg.1 | ENST00000437795.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200006721.hg.1 | ENST00000453100.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200006721.hg.1 | ENST00000453100.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200006721.hg.1 | ENST00000453100.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200006721.hg.1 | ENST00000453100.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200006721.hg.1 | ENST00000453100.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200006721.hg.1 | NR_047499.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200006721.hg.1 | NR_047499.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200006721.hg.1 | NR_047499.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200006721.hg.1 | NR_047499.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200006721.hg.1 | NR_047499.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200006721.hg.1 | NR_047499.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200006721.hg.1 | NR_047499.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200006721.hg.1 | NR_047499.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100008514.hg.1 | ENST00000333090.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008514.hg.1 | ENST00000333090.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008514.hg.1 | ENST00000333090.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008514.hg.1 | ENST00000333090.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008514.hg.1 | ENST00000333090.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008514.hg.1 | ENST00000333090.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008514.hg.1 | ENST00000525167.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008514.hg.1 | ENST00000525167.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008514.hg.1 | ENST00000527881.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008514.hg.1 | ENST00000527881.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008514.hg.1 | ENST00000527881.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008514.hg.1 | ENST00000527881.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008514.hg.1 | ENST00000527881.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008514.hg.1 | ENST00000533752.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008514.hg.1 | ENST00000533752.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008514.hg.1 | ENST00000533752.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008514.hg.1 | ENST00000533752.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008514.hg.1 | ENST00000612930.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008514.hg.1 | ENST00000612930.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008514.hg.1 | ENST00000612930.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008514.hg.1 | ENST00000612930.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008514.hg.1 | ENST00000612930.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008514.hg.1 | NM_001258210.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100008514.hg.1 | NM_001258210.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100008514.hg.1 | NM_001258210.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100008514.hg.1 | NM_001258210.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100008514.hg.1 | NM_001258210.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100008514.hg.1 | NM_015516.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100008514.hg.1 | NM_015516.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100008514.hg.1 | NM_015516.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100008514.hg.1 | NM_015516.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100008514.hg.1 | NM_015516.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100008514.hg.1 | NM_015516.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200006721.hg.1 | ENST00000417473.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200006721.hg.1 | ENST00000417473.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200006721.hg.1 | ENST00000417473.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200006721.hg.1 | ENST00000417473.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200006721.hg.1 | ENST00000417473.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200006721.hg.1 | ENST00000417473.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200006721.hg.1 | ENST00000417473.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200006721.hg.1 | ENST00000417473.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200006721.hg.1 | ENST00000417473.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200006721.hg.1 | ENST00000417473.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200006721.hg.1 | ENST00000417473.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200006721.hg.1 | ENST00000417473.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200006721.hg.1 | ENST00000417473.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200006721.hg.1 | ENST00000417473.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200006721.hg.1 | ENST00000417473.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200006721.hg.1 | ENST00000417473.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200006721.hg.1 | ENST00000432665.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200006721.hg.1 | ENST00000432665.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200006721.hg.1 | ENST00000432665.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200006721.hg.1 | ENST00000432665.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200006721.hg.1 | ENST00000432665.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200006721.hg.1 | ENST00000437795.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200006721.hg.1 | ENST00000437795.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200006721.hg.1 | ENST00000437795.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200006721.hg.1 | ENST00000437795.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200006721.hg.1 | ENST00000437795.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200006721.hg.1 | ENST00000437795.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200006721.hg.1 | ENST00000437795.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200006721.hg.1 | ENST00000437795.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200006721.hg.1 | ENST00000437795.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200006721.hg.1 | ENST00000437795.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200006721.hg.1 | ENST00000437795.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200006721.hg.1 | ENST00000437795.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200006721.hg.1 | ENST00000453100.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200006721.hg.1 | ENST00000453100.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200006721.hg.1 | ENST00000453100.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200006721.hg.1 | ENST00000453100.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200006721.hg.1 | ENST00000453100.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200006721.hg.1 | NR_047499.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200006721.hg.1 | NR_047499.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200006721.hg.1 | NR_047499.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200006721.hg.1 | NR_047499.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200006721.hg.1 | NR_047499.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200006721.hg.1 | NR_047499.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200006721.hg.1 | NR_047499.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200006721.hg.1 | NR_047499.1 | Refseq |

TSKU
Genomic Map