| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200007035.hg.1 |
ENST00000330029.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200007035.hg.1 |
ENST00000330029.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200007035.hg.1 |
ENST00000330029.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200007035.hg.1 |
ENST00000330029.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200007035.hg.1 |
ENST00000330029.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200007035.hg.1 |
ENST00000401406.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200007035.hg.1 |
ENST00000401406.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200007035.hg.1 |
ENST00000401406.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200007035.hg.1 |
ENST00000401406.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200007035.hg.1 |
ENST00000401406.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200007035.hg.1 |
ENST00000616603.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200007035.hg.1 |
ENST00000616603.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200007035.hg.1 |
ENST00000616603.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200007035.hg.1 |
ENST00000616603.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200007035.hg.1 |
ENST00000616603.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200007035.hg.1 |
NM_001003684.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200007035.hg.1 |
NM_001003684.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200007035.hg.1 |
NM_001003684.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200007035.hg.1 |
NM_001003684.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200007035.hg.1 |
NM_001003684.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200007035.hg.1 |
NM_001003684.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200007035.hg.1 |
NM_013387.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200007035.hg.1 |
NM_013387.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200007035.hg.1 |
NM_013387.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200007035.hg.1 |
NM_013387.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200007035.hg.1 |
NM_013387.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2200007035.hg.1 |
ENST00000330029.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200007035.hg.1 |
ENST00000330029.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200007035.hg.1 |
ENST00000330029.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200007035.hg.1 |
ENST00000330029.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200007035.hg.1 |
ENST00000330029.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200007035.hg.1 |
ENST00000401406.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200007035.hg.1 |
ENST00000401406.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200007035.hg.1 |
ENST00000401406.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200007035.hg.1 |
ENST00000401406.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200007035.hg.1 |
ENST00000401406.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200007035.hg.1 |
ENST00000616603.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200007035.hg.1 |
ENST00000616603.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200007035.hg.1 |
ENST00000616603.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200007035.hg.1 |
ENST00000616603.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200007035.hg.1 |
ENST00000616603.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200007035.hg.1 |
NM_001003684.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2200007035.hg.1 |
NM_001003684.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2200007035.hg.1 |
NM_001003684.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2200007035.hg.1 |
NM_001003684.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2200007035.hg.1 |
NM_001003684.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2200007035.hg.1 |
NM_001003684.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2200007035.hg.1 |
NM_013387.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2200007035.hg.1 |
NM_013387.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2200007035.hg.1 |
NM_013387.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2200007035.hg.1 |
NM_013387.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2200007035.hg.1 |
NM_013387.3 |
Refseq |