Assay Details
| Gene Symbol: SEPT5-GP1BB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | BC030503.1 | - | 1 | 496 | 112 | |
| L20860.1 | - | 1 | 112 | |||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

SEPT5-GP1BB| Gene Symbol: SEPT5-GP1BB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | BC030503.1 | - | 1 | 496 | 112 | |
| L20860.1 | - | 1 | 112 | |||
Set Membership: |
|