Assay Details
| Gene Symbol: CAPN2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001146068.1 | NP_001139540.1 | 5 - 6 | 729 | 86 | |
| NM_001748.4 | NP_001739.2 | 5 - 6 | 951 | 86 | ||
| XM_011544286.2 | XP_011542588.1 | 5 - 6 | 1128 | 86 | ||
| GenBank mRNA | AB209855.1 | - | 5 - 6 | 816 | 86 | |
| AF261089.1 | - | 5 - 6 | 854 | 86 | ||
| AK297800.1 | - | 5 - 6 | 729 | 86 | ||
| AK315179.1 | - | 5 - 6 | 853 | 86 | ||
| AK316211.1 | - | 5 - 6 | 772 | 86 | ||
| BC007686.2 | - | 5 - 6 | 951 | 86 | ||
| BC011828.2 | - | 5 - 6 | 951 | 86 | ||
| BC021303.2 | - | 5 - 6 | 845 | 5090865 | 86 | |
| DA448245.1 | - | 2 - 3 | 310 | 86 | ||
| M23254.1 | - | 5 - 6 | 857 | 86 | ||
More Information
Set Membership: |
|

CAPN2
Genomic Map