Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0200011976.hg.1 |
ENST00000313213.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200011976.hg.1 |
ENST00000313213.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200011976.hg.1 |
ENST00000313213.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200011976.hg.1 |
ENST00000313213.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200011976.hg.1 |
ENST00000313213.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200011976.hg.1 |
ENST00000313213.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200011976.hg.1 |
ENST00000313213.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200011976.hg.1 |
ENST00000313213.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200011976.hg.1 |
ENST00000313213.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200011976.hg.1 |
ENST00000436622.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200011976.hg.1 |
ENST00000436622.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200011976.hg.1 |
ENST00000436622.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200011976.hg.1 |
ENST00000436622.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200011976.hg.1 |
ENST00000436622.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200011976.hg.1 |
ENST00000465360.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200011976.hg.1 |
ENST00000465360.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200011976.hg.1 |
ENST00000465360.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200011976.hg.1 |
ENST00000465360.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200011976.hg.1 |
ENST00000465360.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200011976.hg.1 |
ENST00000465360.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200011976.hg.1 |
NM_199346.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200011976.hg.1 |
NM_199346.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200011976.hg.1 |
NM_199346.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200011976.hg.1 |
NM_199346.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200011976.hg.1 |
NM_199346.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200011976.hg.1 |
NM_199346.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200011976.hg.1 |
NM_199346.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200011976.hg.1 |
NM_199346.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200011976.hg.1 |
NM_199346.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200011976.hg.1 |
NM_199346.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200011976.hg.1 |
ENST00000313213.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200011976.hg.1 |
ENST00000313213.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200011976.hg.1 |
ENST00000313213.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200011976.hg.1 |
ENST00000313213.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200011976.hg.1 |
ENST00000313213.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200011976.hg.1 |
ENST00000313213.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200011976.hg.1 |
ENST00000313213.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200011976.hg.1 |
ENST00000313213.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200011976.hg.1 |
ENST00000313213.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200011976.hg.1 |
ENST00000436622.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200011976.hg.1 |
ENST00000436622.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200011976.hg.1 |
ENST00000436622.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200011976.hg.1 |
ENST00000436622.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200011976.hg.1 |
ENST00000436622.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200011976.hg.1 |
ENST00000465360.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200011976.hg.1 |
ENST00000465360.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200011976.hg.1 |
ENST00000465360.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200011976.hg.1 |
ENST00000465360.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200011976.hg.1 |
ENST00000465360.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200011976.hg.1 |
ENST00000465360.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200011976.hg.1 |
NM_199346.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200011976.hg.1 |
NM_199346.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200011976.hg.1 |
NM_199346.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200011976.hg.1 |
NM_199346.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200011976.hg.1 |
NM_199346.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200011976.hg.1 |
NM_199346.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200011976.hg.1 |
NM_199346.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200011976.hg.1 |
NM_199346.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200011976.hg.1 |
NM_199346.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200011976.hg.1 |
NM_199346.2 |
Refseq |