Assay Details
| Gene Symbol: ARSE | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001282628.1 | NP_001269557.1 | 2 - 3 | 271 | 104 | |
| XM_017029526.1 | XP_016885015.1 | 2 - 3 | 316 | 104 | ||
| GenBank mRNA | AK301004.1 | - | 2 - 3 | 102 | 104 | |
| CN270432.1 | - | 2 - 3 | 66 | 104 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK301004.1, NM_001282628.1, XM_017029526.1 transcripts.
Set Membership: |
|
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011339.hg.1 | ENST00000381134.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011339.hg.1 | ENST00000381134.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011339.hg.1 | ENST00000381134.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011339.hg.1 | ENST00000381134.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011339.hg.1 | ENST00000381134.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011339.hg.1 | ENST00000381134.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011339.hg.1 | ENST00000381134.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011339.hg.1 | ENST00000381134.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011339.hg.1 | ENST00000381134.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011339.hg.1 | ENST00000381134.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011339.hg.1 | ENST00000381134.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011339.hg.1 | ENST00000381134.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011339.hg.1 | ENST00000381134.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011339.hg.1 | ENST00000438544.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011339.hg.1 | ENST00000438544.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011339.hg.1 | ENST00000438544.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011339.hg.1 | ENST00000438544.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011339.hg.1 | ENST00000438544.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011339.hg.1 | ENST00000438544.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011339.hg.1 | ENST00000438544.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011339.hg.1 | ENST00000438544.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011339.hg.1 | ENST00000483425.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011339.hg.1 | ENST00000483425.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011339.hg.1 | ENST00000496095.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011339.hg.1 | ENST00000496095.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011339.hg.1 | ENST00000496095.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011339.hg.1 | ENST00000496095.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011339.hg.1 | ENST00000540563.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011339.hg.1 | ENST00000540563.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011339.hg.1 | ENST00000540563.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011339.hg.1 | ENST00000540563.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011339.hg.1 | ENST00000540563.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011339.hg.1 | ENST00000540563.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011339.hg.1 | ENST00000540563.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011339.hg.1 | ENST00000540563.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011339.hg.1 | ENST00000540563.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011339.hg.1 | ENST00000540563.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011339.hg.1 | ENST00000540563.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011339.hg.1 | ENST00000540563.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011339.hg.1 | ENST00000545496.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011339.hg.1 | ENST00000545496.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011339.hg.1 | ENST00000545496.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011339.hg.1 | ENST00000545496.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011339.hg.1 | ENST00000545496.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011339.hg.1 | ENST00000545496.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011339.hg.1 | ENST00000545496.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011339.hg.1 | ENST00000545496.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011339.hg.1 | ENST00000545496.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011339.hg.1 | ENST00000545496.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011339.hg.1 | ENST00000545496.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011339.hg.1 | ENST00000545496.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011339.hg.1 | ENST00000545496.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011339.hg.1 | ENST00000545496.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011339.hg.1 | ENST00000545496.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011339.hg.1 | ENST00000545496.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011339.hg.1 | ENST00000545496.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011339.hg.1 | NM_000047.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011339.hg.1 | NM_000047.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011339.hg.1 | NM_000047.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011339.hg.1 | NM_000047.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011339.hg.1 | NM_000047.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011339.hg.1 | NM_000047.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011339.hg.1 | NM_000047.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011339.hg.1 | NM_000047.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011339.hg.1 | NM_000047.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011339.hg.1 | NM_000047.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011339.hg.1 | NM_000047.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011339.hg.1 | NM_000047.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011339.hg.1 | NM_000047.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011339.hg.1 | NM_000047.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011339.hg.1 | NM_000047.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011339.hg.1 | NM_001282628.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011339.hg.1 | NM_001282628.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011339.hg.1 | NM_001282628.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011339.hg.1 | NM_001282628.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011339.hg.1 | NM_001282628.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011339.hg.1 | NM_001282628.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011339.hg.1 | NM_001282628.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011339.hg.1 | NM_001282628.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011339.hg.1 | NM_001282628.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011339.hg.1 | NM_001282628.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011339.hg.1 | NM_001282628.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011339.hg.1 | NM_001282628.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011339.hg.1 | NM_001282628.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011339.hg.1 | NM_001282628.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011339.hg.1 | NM_001282628.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011339.hg.1 | NM_001282628.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011339.hg.1 | NM_001282628.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011339.hg.1 | NM_001282631.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011339.hg.1 | NM_001282631.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011339.hg.1 | NM_001282631.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011339.hg.1 | NM_001282631.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011339.hg.1 | NM_001282631.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011339.hg.1 | NM_001282631.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011339.hg.1 | NM_001282631.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011339.hg.1 | NM_001282631.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011339.hg.1 | NM_001282631.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011339.hg.1 | NM_001282631.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011339.hg.1 | NM_001282631.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011339.hg.1 | NM_001282631.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00011339.hg.1 | ENST00000381134.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00011339.hg.1 | ENST00000381134.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00011339.hg.1 | ENST00000381134.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00011339.hg.1 | ENST00000381134.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00011339.hg.1 | ENST00000381134.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00011339.hg.1 | ENST00000381134.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00011339.hg.1 | ENST00000381134.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00011339.hg.1 | ENST00000381134.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00011339.hg.1 | ENST00000381134.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00011339.hg.1 | ENST00000381134.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00011339.hg.1 | ENST00000381134.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00011339.hg.1 | ENST00000381134.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00011339.hg.1 | ENST00000381134.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00011339.hg.1 | ENST00000438544.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00011339.hg.1 | ENST00000438544.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00011339.hg.1 | ENST00000438544.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00011339.hg.1 | ENST00000438544.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00011339.hg.1 | ENST00000438544.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00011339.hg.1 | ENST00000438544.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00011339.hg.1 | ENST00000438544.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00011339.hg.1 | ENST00000438544.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00011339.hg.1 | ENST00000483425.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00011339.hg.1 | ENST00000483425.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00011339.hg.1 | ENST00000496095.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00011339.hg.1 | ENST00000496095.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00011339.hg.1 | ENST00000496095.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00011339.hg.1 | ENST00000496095.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00011339.hg.1 | ENST00000540563.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00011339.hg.1 | ENST00000540563.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00011339.hg.1 | ENST00000540563.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00011339.hg.1 | ENST00000540563.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00011339.hg.1 | ENST00000540563.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00011339.hg.1 | ENST00000540563.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00011339.hg.1 | ENST00000540563.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00011339.hg.1 | ENST00000540563.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00011339.hg.1 | ENST00000540563.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00011339.hg.1 | ENST00000540563.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00011339.hg.1 | ENST00000540563.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00011339.hg.1 | ENST00000540563.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00011339.hg.1 | ENST00000545496.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00011339.hg.1 | ENST00000545496.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00011339.hg.1 | ENST00000545496.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00011339.hg.1 | ENST00000545496.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00011339.hg.1 | ENST00000545496.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00011339.hg.1 | ENST00000545496.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00011339.hg.1 | ENST00000545496.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00011339.hg.1 | ENST00000545496.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00011339.hg.1 | ENST00000545496.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00011339.hg.1 | ENST00000545496.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00011339.hg.1 | ENST00000545496.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00011339.hg.1 | ENST00000545496.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00011339.hg.1 | ENST00000545496.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00011339.hg.1 | ENST00000545496.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00011339.hg.1 | ENST00000545496.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00011339.hg.1 | ENST00000545496.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00011339.hg.1 | ENST00000545496.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00011339.hg.1 | NM_000047.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00011339.hg.1 | NM_000047.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00011339.hg.1 | NM_000047.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00011339.hg.1 | NM_000047.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00011339.hg.1 | NM_000047.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00011339.hg.1 | NM_000047.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00011339.hg.1 | NM_000047.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00011339.hg.1 | NM_000047.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00011339.hg.1 | NM_000047.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00011339.hg.1 | NM_000047.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00011339.hg.1 | NM_000047.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00011339.hg.1 | NM_000047.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00011339.hg.1 | NM_000047.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00011339.hg.1 | NM_000047.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00011339.hg.1 | NM_000047.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00011339.hg.1 | NM_001282628.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00011339.hg.1 | NM_001282628.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00011339.hg.1 | NM_001282628.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00011339.hg.1 | NM_001282628.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00011339.hg.1 | NM_001282628.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00011339.hg.1 | NM_001282628.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00011339.hg.1 | NM_001282628.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00011339.hg.1 | NM_001282628.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00011339.hg.1 | NM_001282628.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00011339.hg.1 | NM_001282628.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00011339.hg.1 | NM_001282628.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00011339.hg.1 | NM_001282628.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00011339.hg.1 | NM_001282628.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00011339.hg.1 | NM_001282628.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00011339.hg.1 | NM_001282628.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00011339.hg.1 | NM_001282628.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00011339.hg.1 | NM_001282628.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00011339.hg.1 | NM_001282631.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00011339.hg.1 | NM_001282631.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00011339.hg.1 | NM_001282631.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00011339.hg.1 | NM_001282631.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00011339.hg.1 | NM_001282631.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00011339.hg.1 | NM_001282631.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00011339.hg.1 | NM_001282631.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00011339.hg.1 | NM_001282631.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00011339.hg.1 | NM_001282631.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00011339.hg.1 | NM_001282631.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00011339.hg.1 | NM_001282631.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00011339.hg.1 | NM_001282631.1 | Refseq |

ARSE
Genomic Map