Assay Details
| Gene Symbol: C6orf136 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001109938.2 | NP_001103408.1 | 2 - 3 | 664 | 98 | |
| NM_001161376.1 | NP_001154848.1 | 2 - 3 | 1207 | 98 | ||
| XM_011514384.1 | XP_011512686.1 | 3 - 4 | 756 | 98 | ||
| GenBank mRNA | AK127564.1 | - | 2 - 3 | 624 | 98 | |
| BC016167.1 | - | 2 - 3 | 815 | 3509511 | 98 | |
| BC064397.1 | - | 2 - 3 | 825 | 98 | ||
| BP255323.1 | - | 2 - 3 | 471 | 98 | ||
More Information
Set Membership: |
|

C6orf136
Genomic Map