Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1500008733.hg.1 |
ENST00000566245.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500008733.hg.1 |
ENST00000566245.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500008733.hg.1 |
ENST00000566245.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500008733.hg.1 |
ENST00000567246.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500008733.hg.1 |
ENST00000567246.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500008733.hg.1 |
ENST00000567246.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500008733.hg.1 |
ENST00000567246.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500008733.hg.1 |
ENST00000567854.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500008733.hg.1 |
ENST00000567854.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500008733.hg.1 |
NR_026647.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500008733.hg.1 |
NR_026647.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500008733.hg.1 |
NR_026647.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500008733.hg.1 |
NR_026647.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500008733.hg.1 |
ENST00000566245.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500008733.hg.1 |
ENST00000566245.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500008733.hg.1 |
ENST00000566245.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500008733.hg.1 |
ENST00000567246.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500008733.hg.1 |
ENST00000567246.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500008733.hg.1 |
ENST00000567246.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500008733.hg.1 |
ENST00000567246.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500008733.hg.1 |
ENST00000567854.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500008733.hg.1 |
ENST00000567854.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500008733.hg.1 |
NR_026647.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500008733.hg.1 |
NR_026647.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500008733.hg.1 |
NR_026647.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500008733.hg.1 |
NR_026647.1 |
Refseq |