Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013021.hg.1 |
ENST00000311890.13 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013021.hg.1 |
ENST00000311890.13 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013021.hg.1 |
ENST00000311890.13 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013021.hg.1 |
ENST00000311890.13 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013021.hg.1 |
ENST00000311890.13 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013021.hg.1 |
ENST00000311890.13 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013021.hg.1 |
ENST00000311890.13 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013021.hg.1 |
ENST00000311890.13 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013021.hg.1 |
ENST00000311890.13 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013021.hg.1 |
ENST00000311890.13 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013021.hg.1 |
ENST00000311890.13 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013021.hg.1 |
ENST00000311890.13 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013021.hg.1 |
ENST00000311890.13 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013021.hg.1 |
ENST00000375718.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013021.hg.1 |
ENST00000375718.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013021.hg.1 |
ENST00000375718.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013021.hg.1 |
ENST00000375718.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013021.hg.1 |
ENST00000375718.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013021.hg.1 |
ENST00000375718.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013021.hg.1 |
ENST00000375718.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013021.hg.1 |
ENST00000375718.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013021.hg.1 |
ENST00000406363.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013021.hg.1 |
ENST00000406363.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013021.hg.1 |
ENST00000406363.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013021.hg.1 |
ENST00000406363.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013021.hg.1 |
ENST00000406363.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013021.hg.1 |
ENST00000406363.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013021.hg.1 |
ENST00000406363.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013021.hg.1 |
ENST00000411503.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013021.hg.1 |
ENST00000411503.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013021.hg.1 |
ENST00000411503.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013021.hg.1 |
ENST00000411503.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013021.hg.1 |
ENST00000411503.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013021.hg.1 |
ENST00000411503.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013021.hg.1 |
ENST00000411503.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013021.hg.1 |
ENST00000411503.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013021.hg.1 |
ENST00000411503.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013021.hg.1 |
ENST00000411503.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013021.hg.1 |
ENST00000442459.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013021.hg.1 |
ENST00000442459.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013021.hg.1 |
ENST00000442459.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013021.hg.1 |
ENST00000442459.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013021.hg.1 |
ENST00000442459.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013021.hg.1 |
ENST00000442459.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013021.hg.1 |
ENST00000442459.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013021.hg.1 |
ENST00000442459.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013021.hg.1 |
ENST00000442459.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013021.hg.1 |
ENST00000442459.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013021.hg.1 |
ENST00000442459.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013021.hg.1 |
ENST00000463576.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013021.hg.1 |
ENST00000463576.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013021.hg.1 |
ENST00000463576.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013021.hg.1 |
ENST00000463576.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013021.hg.1 |
ENST00000463576.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013021.hg.1 |
ENST00000487046.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013021.hg.1 |
ENST00000487046.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013021.hg.1 |
ENST00000487046.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013021.hg.1 |
ENST00000487046.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013021.hg.1 |
ENST00000487046.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013021.hg.1 |
ENST00000487046.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013021.hg.1 |
ENST00000487046.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013021.hg.1 |
ENST00000487046.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013021.hg.1 |
ENST00000487046.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013021.hg.1 |
ENST00000487046.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013021.hg.1 |
NM_014424.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013021.hg.1 |
NM_014424.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013021.hg.1 |
NM_014424.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013021.hg.1 |
NM_014424.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013021.hg.1 |
NM_014424.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013021.hg.1 |
NM_014424.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013021.hg.1 |
NM_014424.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013021.hg.1 |
NM_014424.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013021.hg.1 |
NM_014424.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013021.hg.1 |
NM_014424.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013021.hg.1 |
NM_014424.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013021.hg.1 |
NM_014424.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013021.hg.1 |
NM_014424.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100013021.hg.1 |
ENST00000311890.13 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013021.hg.1 |
ENST00000311890.13 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013021.hg.1 |
ENST00000311890.13 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013021.hg.1 |
ENST00000311890.13 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013021.hg.1 |
ENST00000311890.13 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013021.hg.1 |
ENST00000311890.13 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013021.hg.1 |
ENST00000311890.13 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013021.hg.1 |
ENST00000311890.13 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013021.hg.1 |
ENST00000311890.13 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013021.hg.1 |
ENST00000311890.13 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013021.hg.1 |
ENST00000311890.13 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013021.hg.1 |
ENST00000311890.13 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013021.hg.1 |
ENST00000311890.13 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013021.hg.1 |
ENST00000375718.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013021.hg.1 |
ENST00000375718.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013021.hg.1 |
ENST00000375718.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013021.hg.1 |
ENST00000375718.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013021.hg.1 |
ENST00000375718.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013021.hg.1 |
ENST00000375718.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013021.hg.1 |
ENST00000375718.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013021.hg.1 |
ENST00000375718.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013021.hg.1 |
ENST00000406363.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013021.hg.1 |
ENST00000406363.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013021.hg.1 |
ENST00000406363.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013021.hg.1 |
ENST00000406363.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013021.hg.1 |
ENST00000406363.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013021.hg.1 |
ENST00000406363.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013021.hg.1 |
ENST00000406363.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013021.hg.1 |
ENST00000411503.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013021.hg.1 |
ENST00000411503.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013021.hg.1 |
ENST00000411503.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013021.hg.1 |
ENST00000411503.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013021.hg.1 |
ENST00000411503.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013021.hg.1 |
ENST00000411503.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013021.hg.1 |
ENST00000411503.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013021.hg.1 |
ENST00000411503.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013021.hg.1 |
ENST00000411503.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013021.hg.1 |
ENST00000411503.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013021.hg.1 |
ENST00000442459.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013021.hg.1 |
ENST00000442459.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013021.hg.1 |
ENST00000442459.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013021.hg.1 |
ENST00000442459.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013021.hg.1 |
ENST00000442459.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013021.hg.1 |
ENST00000442459.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013021.hg.1 |
ENST00000442459.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013021.hg.1 |
ENST00000442459.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013021.hg.1 |
ENST00000442459.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013021.hg.1 |
ENST00000442459.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013021.hg.1 |
ENST00000442459.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013021.hg.1 |
ENST00000463576.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013021.hg.1 |
ENST00000463576.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013021.hg.1 |
ENST00000463576.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013021.hg.1 |
ENST00000463576.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013021.hg.1 |
ENST00000463576.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013021.hg.1 |
ENST00000487046.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013021.hg.1 |
ENST00000487046.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013021.hg.1 |
ENST00000487046.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013021.hg.1 |
ENST00000487046.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013021.hg.1 |
ENST00000487046.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013021.hg.1 |
ENST00000487046.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013021.hg.1 |
ENST00000487046.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013021.hg.1 |
ENST00000487046.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013021.hg.1 |
ENST00000487046.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013021.hg.1 |
ENST00000487046.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013021.hg.1 |
NM_014424.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100013021.hg.1 |
NM_014424.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100013021.hg.1 |
NM_014424.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100013021.hg.1 |
NM_014424.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100013021.hg.1 |
NM_014424.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100013021.hg.1 |
NM_014424.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100013021.hg.1 |
NM_014424.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100013021.hg.1 |
NM_014424.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100013021.hg.1 |
NM_014424.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100013021.hg.1 |
NM_014424.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100013021.hg.1 |
NM_014424.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100013021.hg.1 |
NM_014424.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100013021.hg.1 |
NM_014424.4 |
Refseq |