Assay Details
| Gene Symbol: CASC2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_026939.1 | 5 | 700 | 94 | ||
| NR_026940.1 | 4 | 600 | 94 | |||
| GenBank mRNA | AJ535621.1 | - | 4 | 413 | 94 | |
| AY226450.1 | - | 5 | 516 | 94 | ||
| BC036393.1 | - | 5 | 701 | 94 | ||
| BC112306.1 | - | 4 | 424 | 94 | ||
| BC112308.1 | - | 4 | 424 | 94 | ||
More Information
Set Membership: |
|

CASC2
Genomic Map