Assay Details
| Gene Symbol: NOD2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AY187242.1 | - | 2 | 90 | 92 | |
More Information
Set Membership: |
|
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1600007827.hg.1 | ENST00000300589.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007827.hg.1 | ENST00000300589.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007827.hg.1 | ENST00000300589.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007827.hg.1 | ENST00000300589.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007827.hg.1 | ENST00000300589.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007827.hg.1 | ENST00000300589.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007827.hg.1 | ENST00000300589.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007827.hg.1 | ENST00000300589.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007827.hg.1 | ENST00000300589.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007827.hg.1 | ENST00000300589.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007827.hg.1 | ENST00000300589.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007827.hg.1 | ENST00000300589.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007827.hg.1 | ENST00000300589.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007827.hg.1 | ENST00000300589.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007827.hg.1 | ENST00000300589.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007827.hg.1 | ENST00000300589.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007827.hg.1 | ENST00000300589.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007827.hg.1 | ENST00000524712.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007827.hg.1 | ENST00000524712.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007827.hg.1 | ENST00000524712.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007827.hg.1 | ENST00000524712.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007827.hg.1 | ENST00000526417.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007827.hg.1 | ENST00000526417.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007827.hg.1 | ENST00000526417.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007827.hg.1 | ENST00000526417.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007827.hg.1 | ENST00000526417.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007827.hg.1 | ENST00000526417.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007827.hg.1 | ENST00000527052.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007827.hg.1 | ENST00000527052.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007827.hg.1 | ENST00000527052.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007827.hg.1 | ENST00000527070.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007827.hg.1 | ENST00000527070.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007827.hg.1 | ENST00000527070.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007827.hg.1 | ENST00000527070.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007827.hg.1 | ENST00000527070.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007827.hg.1 | ENST00000527070.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007827.hg.1 | ENST00000529633.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007827.hg.1 | ENST00000529633.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007827.hg.1 | ENST00000529633.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007827.hg.1 | ENST00000529633.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007827.hg.1 | ENST00000529633.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007827.hg.1 | ENST00000531674.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007827.hg.1 | ENST00000531674.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007827.hg.1 | ENST00000531674.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007827.hg.1 | ENST00000532206.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007827.hg.1 | ENST00000532206.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007827.hg.1 | ENST00000532206.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007827.hg.1 | ENST00000532206.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007827.hg.1 | ENST00000534057.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007827.hg.1 | ENST00000534057.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007827.hg.1 | ENST00000534057.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007827.hg.1 | ENST00000534057.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007827.hg.1 | ENST00000534057.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007827.hg.1 | ENST00000534067.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007827.hg.1 | ENST00000534067.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007827.hg.1 | ENST00000534067.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007827.hg.1 | ENST00000534067.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007827.hg.1 | ENST00000534067.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007827.hg.1 | NM_001293557.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007827.hg.1 | NM_001293557.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007827.hg.1 | NM_001293557.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007827.hg.1 | NM_001293557.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007827.hg.1 | NM_001293557.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007827.hg.1 | NM_001293557.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007827.hg.1 | NM_001293557.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007827.hg.1 | NM_001293557.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007827.hg.1 | NM_001293557.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007827.hg.1 | NM_001293557.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007827.hg.1 | NM_001293557.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007827.hg.1 | NM_001293557.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007827.hg.1 | NM_001293557.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007827.hg.1 | NM_001293557.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007827.hg.1 | NM_001293557.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007827.hg.1 | NM_001293557.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007827.hg.1 | NM_022162.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007827.hg.1 | NM_022162.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007827.hg.1 | NM_022162.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007827.hg.1 | NM_022162.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007827.hg.1 | NM_022162.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007827.hg.1 | NM_022162.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007827.hg.1 | NM_022162.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007827.hg.1 | NM_022162.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007827.hg.1 | NM_022162.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007827.hg.1 | NM_022162.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007827.hg.1 | NM_022162.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007827.hg.1 | NM_022162.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007827.hg.1 | NM_022162.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007827.hg.1 | NM_022162.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007827.hg.1 | NM_022162.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007827.hg.1 | NM_022162.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007827.hg.1 | NM_022162.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600007827.hg.1 | ENST00000300589.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007827.hg.1 | ENST00000300589.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007827.hg.1 | ENST00000300589.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007827.hg.1 | ENST00000300589.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007827.hg.1 | ENST00000300589.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007827.hg.1 | ENST00000300589.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007827.hg.1 | ENST00000300589.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007827.hg.1 | ENST00000300589.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007827.hg.1 | ENST00000300589.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007827.hg.1 | ENST00000300589.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007827.hg.1 | ENST00000300589.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007827.hg.1 | ENST00000300589.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007827.hg.1 | ENST00000300589.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007827.hg.1 | ENST00000300589.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007827.hg.1 | ENST00000300589.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007827.hg.1 | ENST00000300589.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007827.hg.1 | ENST00000300589.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007827.hg.1 | ENST00000524712.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007827.hg.1 | ENST00000524712.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007827.hg.1 | ENST00000524712.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007827.hg.1 | ENST00000524712.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007827.hg.1 | ENST00000526417.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007827.hg.1 | ENST00000526417.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007827.hg.1 | ENST00000526417.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007827.hg.1 | ENST00000526417.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007827.hg.1 | ENST00000526417.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007827.hg.1 | ENST00000526417.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007827.hg.1 | ENST00000527052.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007827.hg.1 | ENST00000527052.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007827.hg.1 | ENST00000527052.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007827.hg.1 | ENST00000527070.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007827.hg.1 | ENST00000527070.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007827.hg.1 | ENST00000527070.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007827.hg.1 | ENST00000527070.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007827.hg.1 | ENST00000527070.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007827.hg.1 | ENST00000527070.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007827.hg.1 | ENST00000529633.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007827.hg.1 | ENST00000529633.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007827.hg.1 | ENST00000529633.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007827.hg.1 | ENST00000529633.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007827.hg.1 | ENST00000529633.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007827.hg.1 | ENST00000531674.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007827.hg.1 | ENST00000531674.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007827.hg.1 | ENST00000531674.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007827.hg.1 | ENST00000532206.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007827.hg.1 | ENST00000532206.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007827.hg.1 | ENST00000532206.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007827.hg.1 | ENST00000532206.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007827.hg.1 | ENST00000534057.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007827.hg.1 | ENST00000534057.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007827.hg.1 | ENST00000534057.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007827.hg.1 | ENST00000534057.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007827.hg.1 | ENST00000534057.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007827.hg.1 | ENST00000534067.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007827.hg.1 | ENST00000534067.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007827.hg.1 | ENST00000534067.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007827.hg.1 | ENST00000534067.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007827.hg.1 | ENST00000534067.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007827.hg.1 | NM_001293557.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600007827.hg.1 | NM_001293557.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600007827.hg.1 | NM_001293557.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600007827.hg.1 | NM_001293557.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600007827.hg.1 | NM_001293557.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600007827.hg.1 | NM_001293557.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600007827.hg.1 | NM_001293557.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600007827.hg.1 | NM_001293557.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600007827.hg.1 | NM_001293557.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600007827.hg.1 | NM_001293557.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600007827.hg.1 | NM_001293557.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600007827.hg.1 | NM_001293557.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600007827.hg.1 | NM_001293557.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600007827.hg.1 | NM_001293557.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600007827.hg.1 | NM_001293557.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600007827.hg.1 | NM_001293557.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600007827.hg.1 | NM_022162.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600007827.hg.1 | NM_022162.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600007827.hg.1 | NM_022162.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600007827.hg.1 | NM_022162.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600007827.hg.1 | NM_022162.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600007827.hg.1 | NM_022162.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600007827.hg.1 | NM_022162.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600007827.hg.1 | NM_022162.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600007827.hg.1 | NM_022162.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600007827.hg.1 | NM_022162.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600007827.hg.1 | NM_022162.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600007827.hg.1 | NM_022162.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600007827.hg.1 | NM_022162.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600007827.hg.1 | NM_022162.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600007827.hg.1 | NM_022162.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600007827.hg.1 | NM_022162.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600007827.hg.1 | NM_022162.2 | Refseq |

NOD2
Genomic Map