Assay Details
| Gene Symbol: GBP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AB208912.1 | - | 7 | 3129 | 85 | |
More Information
Set Membership: |
|
Panther Classification:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0100014852.hg.1 | ENST00000370473.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014852.hg.1 | ENST00000370473.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014852.hg.1 | ENST00000370473.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014852.hg.1 | ENST00000370473.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014852.hg.1 | ENST00000370473.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014852.hg.1 | ENST00000370473.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014852.hg.1 | ENST00000370473.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014852.hg.1 | ENST00000370473.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014852.hg.1 | ENST00000370473.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014852.hg.1 | ENST00000370473.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014852.hg.1 | ENST00000370473.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014852.hg.1 | ENST00000370473.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014852.hg.1 | ENST00000370473.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014852.hg.1 | ENST00000370473.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014852.hg.1 | ENST00000370473.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014852.hg.1 | ENST00000370473.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014852.hg.1 | ENST00000370473.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014852.hg.1 | ENST00000370473.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014852.hg.1 | ENST00000370473.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014852.hg.1 | ENST00000370473.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014852.hg.1 | ENST00000370473.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014852.hg.1 | ENST00000459831.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014852.hg.1 | ENST00000459831.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014852.hg.1 | ENST00000459831.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014852.hg.1 | ENST00000459831.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014852.hg.1 | ENST00000468959.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014852.hg.1 | ENST00000468959.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014852.hg.1 | ENST00000468959.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014852.hg.1 | ENST00000468959.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014852.hg.1 | ENST00000468959.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014852.hg.1 | ENST00000469194.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014852.hg.1 | ENST00000469194.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014852.hg.1 | ENST00000469194.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014852.hg.1 | ENST00000479889.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014852.hg.1 | ENST00000479889.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014852.hg.1 | ENST00000479889.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014852.hg.1 | ENST00000479889.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014852.hg.1 | ENST00000479889.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014852.hg.1 | ENST00000479889.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014852.hg.1 | ENST00000484970.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014852.hg.1 | ENST00000484970.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014852.hg.1 | ENST00000484970.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014852.hg.1 | ENST00000484970.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014852.hg.1 | ENST00000484970.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014852.hg.1 | ENST00000484970.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014852.hg.1 | ENST00000484970.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014852.hg.1 | ENST00000493139.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014852.hg.1 | ENST00000493139.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014852.hg.1 | ENST00000493139.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014852.hg.1 | ENST00000493139.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014852.hg.1 | ENST00000493139.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014852.hg.1 | ENST00000495131.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014852.hg.1 | ENST00000495131.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014852.hg.1 | ENST00000495131.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014852.hg.1 | ENST00000495131.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014852.hg.1 | NM_002053.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014852.hg.1 | NM_002053.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014852.hg.1 | NM_002053.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014852.hg.1 | NM_002053.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014852.hg.1 | NM_002053.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014852.hg.1 | NM_002053.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014852.hg.1 | NM_002053.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014852.hg.1 | NM_002053.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014852.hg.1 | NM_002053.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014852.hg.1 | NM_002053.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014852.hg.1 | NM_002053.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014852.hg.1 | NM_002053.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014852.hg.1 | NM_002053.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014852.hg.1 | NM_002053.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014852.hg.1 | NM_002053.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014852.hg.1 | NM_002053.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014852.hg.1 | NM_002053.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014852.hg.1 | NM_002053.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014852.hg.1 | NM_002053.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014852.hg.1 | NM_002053.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014852.hg.1 | NM_002053.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100014852.hg.1 | ENST00000370473.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014852.hg.1 | ENST00000370473.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014852.hg.1 | ENST00000370473.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014852.hg.1 | ENST00000370473.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014852.hg.1 | ENST00000370473.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014852.hg.1 | ENST00000370473.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014852.hg.1 | ENST00000370473.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014852.hg.1 | ENST00000370473.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014852.hg.1 | ENST00000370473.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014852.hg.1 | ENST00000370473.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014852.hg.1 | ENST00000370473.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014852.hg.1 | ENST00000370473.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014852.hg.1 | ENST00000370473.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014852.hg.1 | ENST00000370473.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014852.hg.1 | ENST00000370473.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014852.hg.1 | ENST00000370473.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014852.hg.1 | ENST00000370473.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014852.hg.1 | ENST00000370473.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014852.hg.1 | ENST00000370473.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014852.hg.1 | ENST00000370473.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014852.hg.1 | ENST00000370473.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014852.hg.1 | ENST00000459831.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014852.hg.1 | ENST00000459831.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014852.hg.1 | ENST00000459831.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014852.hg.1 | ENST00000459831.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014852.hg.1 | ENST00000468959.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014852.hg.1 | ENST00000468959.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014852.hg.1 | ENST00000468959.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014852.hg.1 | ENST00000468959.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014852.hg.1 | ENST00000468959.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014852.hg.1 | ENST00000469194.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014852.hg.1 | ENST00000469194.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014852.hg.1 | ENST00000469194.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014852.hg.1 | ENST00000479889.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014852.hg.1 | ENST00000479889.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014852.hg.1 | ENST00000479889.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014852.hg.1 | ENST00000479889.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014852.hg.1 | ENST00000479889.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014852.hg.1 | ENST00000479889.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014852.hg.1 | ENST00000484970.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014852.hg.1 | ENST00000484970.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014852.hg.1 | ENST00000484970.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014852.hg.1 | ENST00000484970.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014852.hg.1 | ENST00000484970.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014852.hg.1 | ENST00000484970.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014852.hg.1 | ENST00000484970.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014852.hg.1 | ENST00000493139.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014852.hg.1 | ENST00000493139.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014852.hg.1 | ENST00000493139.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014852.hg.1 | ENST00000493139.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014852.hg.1 | ENST00000493139.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014852.hg.1 | ENST00000495131.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014852.hg.1 | ENST00000495131.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014852.hg.1 | ENST00000495131.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014852.hg.1 | ENST00000495131.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014852.hg.1 | NM_002053.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100014852.hg.1 | NM_002053.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100014852.hg.1 | NM_002053.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100014852.hg.1 | NM_002053.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100014852.hg.1 | NM_002053.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100014852.hg.1 | NM_002053.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100014852.hg.1 | NM_002053.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100014852.hg.1 | NM_002053.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100014852.hg.1 | NM_002053.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100014852.hg.1 | NM_002053.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100014852.hg.1 | NM_002053.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100014852.hg.1 | NM_002053.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100014852.hg.1 | NM_002053.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100014852.hg.1 | NM_002053.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100014852.hg.1 | NM_002053.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100014852.hg.1 | NM_002053.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100014852.hg.1 | NM_002053.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100014852.hg.1 | NM_002053.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100014852.hg.1 | NM_002053.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100014852.hg.1 | NM_002053.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100014852.hg.1 | NM_002053.2 | Refseq |

GBP1
Genomic Map