Assay Details
| Gene Symbol: ZDHHC11B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017010116.1 | XP_016865605.1 | 1 | 911 | 104 | |
| XM_017010117.1 | XP_016865606.1 | 1 | 911 | 104 | ||
| XM_017010118.1 | XP_016865607.1 | 1 | 911 | 104 | ||
| XM_017010119.1 | XP_016865608.1 | 1 | 911 | 104 | ||
| XM_017010120.1 | XP_016865609.1 | 1 | 911 | 104 | ||
| XM_017010121.1 | XP_016865610.1 | 1 | 911 | 104 | ||
| XM_017010122.1 | XP_016865611.1 | 1 | 911 | 104 | ||
| XM_017010123.1 | XP_016865612.1 | 1 | 911 | 104 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_017010116.1, XM_017010117.1, XM_017010118.1, XM_017010119.1, XM_017010120.1, XM_017010121.1, XM_017010122.1, XM_017010123.1 transcripts.
Set Membership: |
|

ZDHHC11B
Genomic Map