Assay Details
| Gene Symbol: PCSK1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_000439.4 | NP_000430.3 | 14 | 4269 | 93 | |
| NM_001177875.1 | NP_001171346.1 | 14 | 3927 | 93 | ||
| GenBank mRNA | AB208874.1 | - | 14 | 3947 | 93 | |
| BC031211.1 | - | 2 | 1421 | 93 | ||
| X64810.1 | - | 14 | 4219 | 93 | ||
More Information
Set Membership: |
|

PCSK1
Genomic Map