Assay Details
| Gene Symbol: ZBTB22 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001145338.1 | NP_001138810.1 | 2 | 317 | 98 | |
| NM_005453.4 | NP_005444.4 | 2 | 335 | 98 | ||
| GenBank mRNA | AK095978.1 | - | 1 | 201 | 98 | |
| AK315747.1 | - | 2 | 317 | 98 | ||
| BC018541.1 | - | 2 | 317 | 4385720 | 98 | |
| BX459194.2 | - | 2 | 335 | 98 | ||
| DA777388.1 | - | 2 | 334 | 98 | ||
More Information
Set Membership: |
|

ZBTB22
Genomic Map