Assay Details
Gene Symbol: RSBN1L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_198467.2 | NP_940869.2 | 3 | 873 | 179 | |
GenBank mRNA | BC059402.1 | - | 3 | 856 | 30347338 | 179 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC0700008147.hg.1 | ENST00000334955.12 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700008147.hg.1 | ENST00000334955.12 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700008147.hg.1 | ENST00000334955.12 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700008147.hg.1 | ENST00000334955.12 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700008147.hg.1 | ENST00000334955.12 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700008147.hg.1 | ENST00000334955.12 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700008147.hg.1 | ENST00000334955.12 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700008147.hg.1 | ENST00000334955.12 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700008147.hg.1 | ENST00000334955.12 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700008147.hg.1 | ENST00000334955.12 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700008147.hg.1 | ENST00000334955.12 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700008147.hg.1 | ENST00000334955.12 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700008147.hg.1 | ENST00000334955.12 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700008147.hg.1 | ENST00000334955.12 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700008147.hg.1 | ENST00000334955.12 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700008147.hg.1 | ENST00000441514.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700008147.hg.1 | ENST00000441514.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700008147.hg.1 | ENST00000441514.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700008147.hg.1 | ENST00000441514.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700008147.hg.1 | ENST00000445288.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700008147.hg.1 | ENST00000445288.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700008147.hg.1 | ENST00000445288.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700008147.hg.1 | ENST00000445288.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700008147.hg.1 | ENST00000445288.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700008147.hg.1 | ENST00000445288.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700008147.hg.1 | ENST00000445288.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700008147.hg.1 | ENST00000445288.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700008147.hg.1 | ENST00000445288.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700008147.hg.1 | ENST00000445288.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700008147.hg.1 | ENST00000445288.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700008147.hg.1 | ENST00000445512.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700008147.hg.1 | ENST00000445512.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700008147.hg.1 | ENST00000445512.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700008147.hg.1 | ENST00000445512.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700008147.hg.1 | ENST00000445512.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700008147.hg.1 | ENST00000445512.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700008147.hg.1 | ENST00000462800.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700008147.hg.1 | ENST00000462800.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700008147.hg.1 | ENST00000462800.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700008147.hg.1 | ENST00000462800.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700008147.hg.1 | ENST00000468035.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700008147.hg.1 | ENST00000468035.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700008147.hg.1 | ENST00000468035.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700008147.hg.1 | ENST00000468035.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700008147.hg.1 | ENST00000468035.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700008147.hg.1 | ENST00000614027.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700008147.hg.1 | ENST00000614027.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700008147.hg.1 | ENST00000614027.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700008147.hg.1 | ENST00000614027.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700008147.hg.1 | ENST00000614027.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700008147.hg.1 | ENST00000614027.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700008147.hg.1 | ENST00000614027.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700008147.hg.1 | ENST00000614027.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700008147.hg.1 | ENST00000614027.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700008147.hg.1 | ENST00000614027.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700008147.hg.1 | ENST00000614027.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700008147.hg.1 | ENST00000614027.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700008147.hg.1 | NM_198467.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700008147.hg.1 | NM_198467.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700008147.hg.1 | NM_198467.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700008147.hg.1 | NM_198467.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700008147.hg.1 | NM_198467.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700008147.hg.1 | NM_198467.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700008147.hg.1 | NM_198467.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700008147.hg.1 | NM_198467.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700008147.hg.1 | NM_198467.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700008147.hg.1 | NM_198467.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700008147.hg.1 | NM_198467.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700008147.hg.1 | NM_198467.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700008147.hg.1 | NM_198467.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0700008147.hg.1 | ENST00000334955.12 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700008147.hg.1 | ENST00000334955.12 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700008147.hg.1 | ENST00000334955.12 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700008147.hg.1 | ENST00000334955.12 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700008147.hg.1 | ENST00000334955.12 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700008147.hg.1 | ENST00000334955.12 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700008147.hg.1 | ENST00000334955.12 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700008147.hg.1 | ENST00000334955.12 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700008147.hg.1 | ENST00000334955.12 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700008147.hg.1 | ENST00000334955.12 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700008147.hg.1 | ENST00000334955.12 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700008147.hg.1 | ENST00000334955.12 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700008147.hg.1 | ENST00000334955.12 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700008147.hg.1 | ENST00000334955.12 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700008147.hg.1 | ENST00000334955.12 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700008147.hg.1 | ENST00000441514.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700008147.hg.1 | ENST00000441514.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700008147.hg.1 | ENST00000441514.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700008147.hg.1 | ENST00000441514.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700008147.hg.1 | ENST00000445288.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700008147.hg.1 | ENST00000445288.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700008147.hg.1 | ENST00000445288.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700008147.hg.1 | ENST00000445288.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700008147.hg.1 | ENST00000445288.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700008147.hg.1 | ENST00000445288.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700008147.hg.1 | ENST00000445288.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700008147.hg.1 | ENST00000445288.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700008147.hg.1 | ENST00000445288.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700008147.hg.1 | ENST00000445288.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700008147.hg.1 | ENST00000445288.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700008147.hg.1 | ENST00000445512.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700008147.hg.1 | ENST00000445512.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700008147.hg.1 | ENST00000445512.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700008147.hg.1 | ENST00000445512.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700008147.hg.1 | ENST00000445512.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700008147.hg.1 | ENST00000445512.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700008147.hg.1 | ENST00000462800.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700008147.hg.1 | ENST00000462800.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700008147.hg.1 | ENST00000462800.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700008147.hg.1 | ENST00000462800.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700008147.hg.1 | ENST00000468035.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700008147.hg.1 | ENST00000468035.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700008147.hg.1 | ENST00000468035.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700008147.hg.1 | ENST00000468035.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700008147.hg.1 | ENST00000468035.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700008147.hg.1 | ENST00000614027.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700008147.hg.1 | ENST00000614027.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700008147.hg.1 | ENST00000614027.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700008147.hg.1 | ENST00000614027.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700008147.hg.1 | ENST00000614027.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700008147.hg.1 | ENST00000614027.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700008147.hg.1 | ENST00000614027.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700008147.hg.1 | ENST00000614027.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700008147.hg.1 | ENST00000614027.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700008147.hg.1 | ENST00000614027.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700008147.hg.1 | ENST00000614027.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700008147.hg.1 | ENST00000614027.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700008147.hg.1 | NM_198467.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0700008147.hg.1 | NM_198467.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0700008147.hg.1 | NM_198467.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0700008147.hg.1 | NM_198467.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0700008147.hg.1 | NM_198467.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0700008147.hg.1 | NM_198467.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0700008147.hg.1 | NM_198467.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0700008147.hg.1 | NM_198467.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0700008147.hg.1 | NM_198467.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0700008147.hg.1 | NM_198467.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0700008147.hg.1 | NM_198467.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0700008147.hg.1 | NM_198467.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0700008147.hg.1 | NM_198467.2 | Refseq |
