Assay Details
| Gene Symbol: ASH1L-IT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AW340519.1 | - | 2 | 206 | 155 | |
| AW452990.1 | - | 2 | 223 | 155 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

ASH1L-IT1| Gene Symbol: ASH1L-IT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AW340519.1 | - | 2 | 206 | 155 | |
| AW452990.1 | - | 2 | 223 | 155 | ||
Set Membership: |
|