Assay Details
| Gene Symbol: TESMIN | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001039656.1 | NP_001034745.1 | 3 | 691 | 84 | |
| NM_004923.3 | NP_004914.2 | 3 | 691 | 84 | ||
| XM_011545402.1 | XP_011543704.1 | 3 | 735 | 84 | ||
| XM_011545403.2 | XP_011543705.1 | 2 | 604 | 84 | ||
| XM_017018588.1 | XP_016874077.1 | 3 | 735 | 84 | ||
| GenBank mRNA | AK074817.1 | - | 3 | 638 | 84 | |
| AK292358.1 | - | 3 | 631 | 84 | ||
| BC027593.1 | - | 3 | 627 | 4838081 | 84 | |
| BC054047.2 | - | 2 | 502 | 84 | ||
| BC064579.1 | - | 3 | 658 | 5743865 | 84 | |
| U86074.1 | - | 2 | 330 | 84 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the U86074.1 transcripts.
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1100011394.hg.1 | ENST00000255087.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011394.hg.1 | ENST00000255087.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011394.hg.1 | ENST00000255087.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011394.hg.1 | ENST00000255087.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011394.hg.1 | ENST00000255087.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011394.hg.1 | ENST00000255087.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011394.hg.1 | ENST00000255087.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011394.hg.1 | ENST00000255087.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011394.hg.1 | ENST00000255087.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011394.hg.1 | ENST00000255087.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011394.hg.1 | ENST00000255087.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011394.hg.1 | ENST00000255087.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011394.hg.1 | ENST00000255087.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011394.hg.1 | ENST00000255087.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011394.hg.1 | ENST00000255087.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011394.hg.1 | ENST00000255087.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011394.hg.1 | ENST00000255087.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011394.hg.1 | ENST00000255087.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011394.hg.1 | ENST00000255087.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011394.hg.1 | ENST00000255087.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011394.hg.1 | ENST00000432435.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011394.hg.1 | ENST00000432435.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011394.hg.1 | ENST00000432435.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011394.hg.1 | ENST00000432435.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011394.hg.1 | ENST00000432435.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011394.hg.1 | ENST00000438124.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011394.hg.1 | ENST00000438124.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011394.hg.1 | ENST00000443940.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011394.hg.1 | ENST00000443940.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011394.hg.1 | ENST00000443940.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011394.hg.1 | ENST00000443940.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011394.hg.1 | ENST00000443940.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011394.hg.1 | ENST00000443940.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011394.hg.1 | ENST00000443940.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011394.hg.1 | ENST00000443940.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011394.hg.1 | ENST00000443940.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011394.hg.1 | ENST00000443940.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011394.hg.1 | ENST00000443940.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011394.hg.1 | ENST00000443940.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011394.hg.1 | ENST00000443940.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011394.hg.1 | ENST00000443940.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011394.hg.1 | ENST00000538944.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011394.hg.1 | ENST00000538944.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011394.hg.1 | ENST00000538944.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011394.hg.1 | ENST00000540869.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011394.hg.1 | ENST00000540869.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011394.hg.1 | ENST00000540869.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011394.hg.1 | ENST00000540869.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011394.hg.1 | ENST00000540869.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011394.hg.1 | ENST00000540869.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011394.hg.1 | ENST00000543240.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011394.hg.1 | ENST00000543240.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011394.hg.1 | ENST00000543240.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011394.hg.1 | ENST00000543240.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011394.hg.1 | ENST00000543240.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011394.hg.1 | ENST00000544398.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011394.hg.1 | ENST00000544398.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011394.hg.1 | ENST00000544398.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011394.hg.1 | ENST00000544398.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011394.hg.1 | ENST00000544963.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011394.hg.1 | ENST00000544963.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011394.hg.1 | ENST00000544963.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011394.hg.1 | ENST00000544963.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011394.hg.1 | ENST00000544963.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011394.hg.1 | ENST00000544963.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011394.hg.1 | ENST00000544963.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011394.hg.1 | ENST00000544963.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011394.hg.1 | ENST00000544963.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011394.hg.1 | ENST00000544963.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011394.hg.1 | ENST00000544963.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011394.hg.1 | ENST00000544963.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011394.hg.1 | NM_001039656.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011394.hg.1 | NM_001039656.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011394.hg.1 | NM_001039656.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011394.hg.1 | NM_001039656.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011394.hg.1 | NM_001039656.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011394.hg.1 | NM_001039656.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011394.hg.1 | NM_001039656.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011394.hg.1 | NM_001039656.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011394.hg.1 | NM_001039656.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011394.hg.1 | NM_001039656.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011394.hg.1 | NM_001039656.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011394.hg.1 | NM_001039656.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011394.hg.1 | NM_001039656.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011394.hg.1 | NM_001039656.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011394.hg.1 | NM_001039656.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011394.hg.1 | NM_001039656.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011394.hg.1 | NM_004923.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011394.hg.1 | NM_004923.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011394.hg.1 | NM_004923.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011394.hg.1 | NM_004923.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011394.hg.1 | NM_004923.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011394.hg.1 | NM_004923.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011394.hg.1 | NM_004923.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011394.hg.1 | NM_004923.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011394.hg.1 | NM_004923.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011394.hg.1 | NM_004923.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011394.hg.1 | NM_004923.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011394.hg.1 | NM_004923.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011394.hg.1 | NM_004923.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011394.hg.1 | NM_004923.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011394.hg.1 | NM_004923.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011394.hg.1 | NM_004923.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011394.hg.1 | NM_004923.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011394.hg.1 | NM_004923.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011394.hg.1 | NM_004923.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100011394.hg.1 | ENST00000255087.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011394.hg.1 | ENST00000255087.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011394.hg.1 | ENST00000255087.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011394.hg.1 | ENST00000255087.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011394.hg.1 | ENST00000255087.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011394.hg.1 | ENST00000255087.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011394.hg.1 | ENST00000255087.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011394.hg.1 | ENST00000255087.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011394.hg.1 | ENST00000255087.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011394.hg.1 | ENST00000255087.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011394.hg.1 | ENST00000255087.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011394.hg.1 | ENST00000255087.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011394.hg.1 | ENST00000255087.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011394.hg.1 | ENST00000255087.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011394.hg.1 | ENST00000255087.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011394.hg.1 | ENST00000255087.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011394.hg.1 | ENST00000255087.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011394.hg.1 | ENST00000255087.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011394.hg.1 | ENST00000255087.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011394.hg.1 | ENST00000255087.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011394.hg.1 | ENST00000432435.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011394.hg.1 | ENST00000432435.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011394.hg.1 | ENST00000432435.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011394.hg.1 | ENST00000432435.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011394.hg.1 | ENST00000432435.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011394.hg.1 | ENST00000438124.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011394.hg.1 | ENST00000438124.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011394.hg.1 | ENST00000443940.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011394.hg.1 | ENST00000443940.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011394.hg.1 | ENST00000443940.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011394.hg.1 | ENST00000443940.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011394.hg.1 | ENST00000443940.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011394.hg.1 | ENST00000443940.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011394.hg.1 | ENST00000443940.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011394.hg.1 | ENST00000443940.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011394.hg.1 | ENST00000443940.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011394.hg.1 | ENST00000443940.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011394.hg.1 | ENST00000443940.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011394.hg.1 | ENST00000443940.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011394.hg.1 | ENST00000443940.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011394.hg.1 | ENST00000443940.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011394.hg.1 | ENST00000538944.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011394.hg.1 | ENST00000538944.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011394.hg.1 | ENST00000538944.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011394.hg.1 | ENST00000540869.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011394.hg.1 | ENST00000540869.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011394.hg.1 | ENST00000540869.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011394.hg.1 | ENST00000540869.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011394.hg.1 | ENST00000540869.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011394.hg.1 | ENST00000540869.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011394.hg.1 | ENST00000543240.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011394.hg.1 | ENST00000543240.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011394.hg.1 | ENST00000543240.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011394.hg.1 | ENST00000543240.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011394.hg.1 | ENST00000543240.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011394.hg.1 | ENST00000544398.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011394.hg.1 | ENST00000544398.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011394.hg.1 | ENST00000544398.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011394.hg.1 | ENST00000544398.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011394.hg.1 | ENST00000544963.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011394.hg.1 | ENST00000544963.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011394.hg.1 | ENST00000544963.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011394.hg.1 | ENST00000544963.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011394.hg.1 | ENST00000544963.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011394.hg.1 | ENST00000544963.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011394.hg.1 | ENST00000544963.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011394.hg.1 | ENST00000544963.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011394.hg.1 | ENST00000544963.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011394.hg.1 | ENST00000544963.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011394.hg.1 | ENST00000544963.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011394.hg.1 | ENST00000544963.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011394.hg.1 | NM_001039656.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100011394.hg.1 | NM_001039656.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100011394.hg.1 | NM_001039656.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100011394.hg.1 | NM_001039656.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100011394.hg.1 | NM_001039656.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100011394.hg.1 | NM_001039656.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100011394.hg.1 | NM_001039656.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100011394.hg.1 | NM_001039656.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100011394.hg.1 | NM_001039656.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100011394.hg.1 | NM_001039656.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100011394.hg.1 | NM_001039656.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100011394.hg.1 | NM_001039656.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100011394.hg.1 | NM_001039656.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100011394.hg.1 | NM_001039656.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100011394.hg.1 | NM_001039656.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100011394.hg.1 | NM_001039656.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100011394.hg.1 | NM_004923.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100011394.hg.1 | NM_004923.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100011394.hg.1 | NM_004923.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100011394.hg.1 | NM_004923.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100011394.hg.1 | NM_004923.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100011394.hg.1 | NM_004923.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100011394.hg.1 | NM_004923.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100011394.hg.1 | NM_004923.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100011394.hg.1 | NM_004923.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100011394.hg.1 | NM_004923.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100011394.hg.1 | NM_004923.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100011394.hg.1 | NM_004923.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100011394.hg.1 | NM_004923.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100011394.hg.1 | NM_004923.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100011394.hg.1 | NM_004923.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100011394.hg.1 | NM_004923.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100011394.hg.1 | NM_004923.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100011394.hg.1 | NM_004923.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100011394.hg.1 | NM_004923.3 | Refseq |

TESMIN
Genomic Map