Assay Details
| Gene Symbol: FSCN3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_020369.2 | NP_065102.1 | 2 | 854 | 196 | |
| GenBank mRNA | AF281049.1 | - | 2 | 854 | 196 | |
| AK225867.1 | - | 2 | 808 | 196 | ||
| AK300518.1 | - | 3 | 898 | 196 | ||
| AK314050.1 | - | 2 | 759 | 196 | ||
| BC035606.1 | - | 2 | 729 | 5172588 | 196 | |
More Information
Set Membership: |
|

FSCN3
Genomic Map