Assay Details
| Gene Symbol: VCPIP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_025054.4 | NP_079330.2 | 3 | 7287 | 187 | |
| GenBank mRNA | AK297345.1 | - | 2 | 2920 | 187 | |
| BC049379.1 | - | 3 | 5706 | 187 | ||
| CR936659.1 | - | 3 | 7316 | 187 | ||
More Information
Set Membership: |
|

VCPIP1
Genomic Map