Assay Details
| Gene Symbol: GALNT2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001291866.1 | NP_001278795.1 | 16 | 2836 | 145 | |
| NM_004481.4 | NP_004472.1 | 16 | 2758 | 145 | ||
| GenBank mRNA | AK056187.1 | - | 1 | 742 | 145 | |
| AK098468.1 | - | 1 | 380 | 145 | ||
| BC041120.1 | - | 16 | 2744 | 5553465 | 145 | |
| BC050583.1 | - | 6 | 2324 | 145 | ||
More Information
Set Membership: |
|

GALNT2
Genomic Map