Assay Details
| Gene Symbol: CACFD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_011518181.2 | XP_011516483.1 | 1 | 157 | 84 | |
| XM_011518185.2 | XP_011516487.1 | 1 | 157 | 84 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_011518181.2, XM_011518185.2 transcripts.
Set Membership: |
|

CACFD1
Genomic Map