Assay Details
| Gene Symbol: USP30-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_038996.1 | 2 | 448 | 81 | ||
| GenBank mRNA | AB371497.1 | - | 2 | 435 | 81 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

USP30-AS1| Gene Symbol: USP30-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_038996.1 | 2 | 448 | 81 | ||
| GenBank mRNA | AB371497.1 | - | 2 | 435 | 81 | |
Set Membership: |
|