Assay Details
| Gene Symbol: STS | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001320750.1 | NP_001307679.1 | 2 | 389 | 64 | |
| NM_001320751.1 | NP_001307680.1 | 3 | 383 | 64 | ||
| NM_001320752.1 | NP_001307681.1 | 2 | 229 | 64 | ||
| GenBank mRNA | AM072428.1 | - | 2 | 115 | 64 | |
| AM072429.1 | - | 2 | 138 | 64 | ||
| AM282551.1 | - | 2 | 388 | 64 | ||
| AM282552.1 | - | 2 | 379 | 64 | ||
| DA327298.1 | - | 2 | 192 | 64 | ||
| DA406759.1 | - | 2 | 230 | 64 | ||
| DQ851171.1 | - | 1 | 38 | 64 | ||
| FM209047.1 | - | 3 | 234 | 64 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AM072428.1, AM072429.1, AM282551.1, AM282552.1, DQ851171.1, FM209047.1, NM_001320750.1, NM_001320751.1, NM_001320752.1 transcripts.
Set Membership: |
|

STS
Genomic Map