Assay Details
| Gene Symbol: ARSE | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_000047.2 | NP_000038.2 | 11 | 1846 | 68 | |
| NM_001282628.1 | NP_001269557.1 | 12 | 2132 | 68 | ||
| NM_001282631.1 | NP_001269560.1 | 10 | 1616 | 68 | ||
| XM_005274518.2 | XP_005274575.1 | 11 | 1773 | 68 | ||
| XM_005274519.4 | XP_005274576.1 | 11 | 1936 | 68 | ||
| XM_005274521.4 | XP_005274578.1 | 10 | 1729 | 68 | ||
| XM_011545521.1 | XP_011543823.1 | 9 | 1416 | 68 | ||
| XM_017029525.1 | XP_016885014.1 | 12 | 2063 | 68 | ||
| XM_017029526.1 | XP_016885015.1 | 10 | 1616 | 68 | ||
| GenBank mRNA | AK223183.1 | - | 11 | 1676 | 68 | |
| AK223199.1 | - | 11 | 1643 | 68 | ||
| AK293648.1 | - | 11 | 1551 | 68 | ||
| AK301004.1 | - | 12 | 1963 | 68 | ||
| AK313093.1 | - | 11 | 1665 | 68 | ||
| BC130438.1 | - | 11 | 1641 | 40146469 | 68 | |
| X83573.1 | - | 11 | 1663 | 68 | ||
More Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0Y00006625.hg.1 | ENST00000420443.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0Y00006625.hg.1 | ENST00000420443.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0Y00006625.hg.1 | ENST00000420443.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0Y00006625.hg.1 | ENST00000430152.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0Y00006625.hg.1 | ENST00000430152.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011339.hg.1 | ENST00000381134.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011339.hg.1 | ENST00000381134.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011339.hg.1 | ENST00000381134.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011339.hg.1 | ENST00000381134.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011339.hg.1 | ENST00000381134.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011339.hg.1 | ENST00000381134.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011339.hg.1 | ENST00000381134.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011339.hg.1 | ENST00000381134.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011339.hg.1 | ENST00000381134.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011339.hg.1 | ENST00000381134.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011339.hg.1 | ENST00000381134.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011339.hg.1 | ENST00000381134.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011339.hg.1 | ENST00000381134.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011339.hg.1 | ENST00000438544.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011339.hg.1 | ENST00000438544.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011339.hg.1 | ENST00000438544.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011339.hg.1 | ENST00000438544.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011339.hg.1 | ENST00000438544.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011339.hg.1 | ENST00000438544.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011339.hg.1 | ENST00000438544.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011339.hg.1 | ENST00000438544.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011339.hg.1 | ENST00000483425.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011339.hg.1 | ENST00000483425.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011339.hg.1 | ENST00000496095.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011339.hg.1 | ENST00000496095.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011339.hg.1 | ENST00000496095.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011339.hg.1 | ENST00000496095.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011339.hg.1 | ENST00000540563.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011339.hg.1 | ENST00000540563.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011339.hg.1 | ENST00000540563.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011339.hg.1 | ENST00000540563.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011339.hg.1 | ENST00000540563.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011339.hg.1 | ENST00000540563.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011339.hg.1 | ENST00000540563.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011339.hg.1 | ENST00000540563.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011339.hg.1 | ENST00000540563.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011339.hg.1 | ENST00000540563.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011339.hg.1 | ENST00000540563.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011339.hg.1 | ENST00000540563.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011339.hg.1 | ENST00000545496.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011339.hg.1 | ENST00000545496.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011339.hg.1 | ENST00000545496.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011339.hg.1 | ENST00000545496.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011339.hg.1 | ENST00000545496.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011339.hg.1 | ENST00000545496.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011339.hg.1 | ENST00000545496.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011339.hg.1 | ENST00000545496.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011339.hg.1 | ENST00000545496.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011339.hg.1 | ENST00000545496.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011339.hg.1 | ENST00000545496.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011339.hg.1 | ENST00000545496.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011339.hg.1 | ENST00000545496.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011339.hg.1 | ENST00000545496.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011339.hg.1 | ENST00000545496.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011339.hg.1 | ENST00000545496.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011339.hg.1 | ENST00000545496.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011339.hg.1 | NM_000047.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011339.hg.1 | NM_000047.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011339.hg.1 | NM_000047.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011339.hg.1 | NM_000047.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011339.hg.1 | NM_000047.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011339.hg.1 | NM_000047.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011339.hg.1 | NM_000047.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011339.hg.1 | NM_000047.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011339.hg.1 | NM_000047.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011339.hg.1 | NM_000047.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011339.hg.1 | NM_000047.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011339.hg.1 | NM_000047.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011339.hg.1 | NM_000047.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011339.hg.1 | NM_000047.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011339.hg.1 | NM_000047.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011339.hg.1 | NM_001282628.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011339.hg.1 | NM_001282628.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011339.hg.1 | NM_001282628.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011339.hg.1 | NM_001282628.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011339.hg.1 | NM_001282628.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011339.hg.1 | NM_001282628.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011339.hg.1 | NM_001282628.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011339.hg.1 | NM_001282628.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011339.hg.1 | NM_001282628.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011339.hg.1 | NM_001282628.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011339.hg.1 | NM_001282628.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011339.hg.1 | NM_001282628.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011339.hg.1 | NM_001282628.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011339.hg.1 | NM_001282628.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011339.hg.1 | NM_001282628.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011339.hg.1 | NM_001282628.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011339.hg.1 | NM_001282628.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011339.hg.1 | NM_001282631.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011339.hg.1 | NM_001282631.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011339.hg.1 | NM_001282631.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011339.hg.1 | NM_001282631.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011339.hg.1 | NM_001282631.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011339.hg.1 | NM_001282631.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011339.hg.1 | NM_001282631.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011339.hg.1 | NM_001282631.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011339.hg.1 | NM_001282631.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011339.hg.1 | NM_001282631.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011339.hg.1 | NM_001282631.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011339.hg.1 | NM_001282631.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00011339.hg.1 | ENST00000381134.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00011339.hg.1 | ENST00000381134.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00011339.hg.1 | ENST00000381134.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00011339.hg.1 | ENST00000381134.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00011339.hg.1 | ENST00000381134.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00011339.hg.1 | ENST00000381134.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00011339.hg.1 | ENST00000381134.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00011339.hg.1 | ENST00000381134.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00011339.hg.1 | ENST00000381134.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00011339.hg.1 | ENST00000381134.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00011339.hg.1 | ENST00000381134.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00011339.hg.1 | ENST00000381134.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00011339.hg.1 | ENST00000381134.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00011339.hg.1 | ENST00000438544.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00011339.hg.1 | ENST00000438544.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00011339.hg.1 | ENST00000438544.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00011339.hg.1 | ENST00000438544.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00011339.hg.1 | ENST00000438544.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00011339.hg.1 | ENST00000438544.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00011339.hg.1 | ENST00000438544.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00011339.hg.1 | ENST00000438544.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00011339.hg.1 | ENST00000483425.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00011339.hg.1 | ENST00000483425.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00011339.hg.1 | ENST00000496095.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00011339.hg.1 | ENST00000496095.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00011339.hg.1 | ENST00000496095.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00011339.hg.1 | ENST00000496095.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00011339.hg.1 | ENST00000540563.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00011339.hg.1 | ENST00000540563.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00011339.hg.1 | ENST00000540563.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00011339.hg.1 | ENST00000540563.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00011339.hg.1 | ENST00000540563.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00011339.hg.1 | ENST00000540563.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00011339.hg.1 | ENST00000540563.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00011339.hg.1 | ENST00000540563.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00011339.hg.1 | ENST00000540563.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00011339.hg.1 | ENST00000540563.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00011339.hg.1 | ENST00000540563.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00011339.hg.1 | ENST00000540563.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00011339.hg.1 | ENST00000545496.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00011339.hg.1 | ENST00000545496.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00011339.hg.1 | ENST00000545496.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00011339.hg.1 | ENST00000545496.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00011339.hg.1 | ENST00000545496.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00011339.hg.1 | ENST00000545496.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00011339.hg.1 | ENST00000545496.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00011339.hg.1 | ENST00000545496.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00011339.hg.1 | ENST00000545496.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00011339.hg.1 | ENST00000545496.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00011339.hg.1 | ENST00000545496.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00011339.hg.1 | ENST00000545496.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00011339.hg.1 | ENST00000545496.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00011339.hg.1 | ENST00000545496.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00011339.hg.1 | ENST00000545496.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00011339.hg.1 | ENST00000545496.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00011339.hg.1 | ENST00000545496.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00011339.hg.1 | NM_000047.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00011339.hg.1 | NM_000047.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00011339.hg.1 | NM_000047.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00011339.hg.1 | NM_000047.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00011339.hg.1 | NM_000047.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00011339.hg.1 | NM_000047.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00011339.hg.1 | NM_000047.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00011339.hg.1 | NM_000047.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00011339.hg.1 | NM_000047.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00011339.hg.1 | NM_000047.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00011339.hg.1 | NM_000047.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00011339.hg.1 | NM_000047.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00011339.hg.1 | NM_000047.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00011339.hg.1 | NM_000047.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00011339.hg.1 | NM_000047.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00011339.hg.1 | NM_001282628.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00011339.hg.1 | NM_001282628.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00011339.hg.1 | NM_001282628.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00011339.hg.1 | NM_001282628.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00011339.hg.1 | NM_001282628.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00011339.hg.1 | NM_001282628.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00011339.hg.1 | NM_001282628.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00011339.hg.1 | NM_001282628.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00011339.hg.1 | NM_001282628.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00011339.hg.1 | NM_001282628.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00011339.hg.1 | NM_001282628.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00011339.hg.1 | NM_001282628.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00011339.hg.1 | NM_001282628.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00011339.hg.1 | NM_001282628.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00011339.hg.1 | NM_001282628.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00011339.hg.1 | NM_001282628.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00011339.hg.1 | NM_001282628.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00011339.hg.1 | NM_001282631.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00011339.hg.1 | NM_001282631.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00011339.hg.1 | NM_001282631.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00011339.hg.1 | NM_001282631.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00011339.hg.1 | NM_001282631.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00011339.hg.1 | NM_001282631.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00011339.hg.1 | NM_001282631.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00011339.hg.1 | NM_001282631.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00011339.hg.1 | NM_001282631.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00011339.hg.1 | NM_001282631.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00011339.hg.1 | NM_001282631.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00011339.hg.1 | NM_001282631.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0Y00006625.hg.1 | ENST00000420443.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0Y00006625.hg.1 | ENST00000420443.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0Y00006625.hg.1 | ENST00000420443.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0Y00006625.hg.1 | ENST00000430152.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0Y00006625.hg.1 | ENST00000430152.1 | Ensembl |

ARSE
Genomic Map