Assay Details
| Gene Symbol: LINC01556 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_103538.1 | 1 | 503 | 104 | ||
| GenBank mRNA | BX118328.1 | - | 1 | 503 | 104 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LINC01556| Gene Symbol: LINC01556 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_103538.1 | 1 | 503 | 104 | ||
| GenBank mRNA | BX118328.1 | - | 1 | 503 | 104 | |
Set Membership: |
|