Assay Details
| Gene Symbol: NEMP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_011538056.2 | XP_011536358.1 | 1 | 179 | 81 | |
| XM_011538058.2 | XP_011536360.1 | 1 | 176 | 81 | ||
| XM_011538060.2 | XP_011536362.1 | 1 | 216 | 81 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_011538056.2, XM_011538058.2, XM_011538060.2 transcripts.
Set Membership: |
|

NEMP1
Genomic Map